Spex Sodium Carbonate (Na2CO3) Eluent Concentrates
Eluent concentrates for ion chromatography
  • Manufactured under ISO 9001, ISO/IEC 17025 and ISO 17034 compliance
  • Made from high-purity salts and filtered ASTM Type I Water
Ion chromatography (IC) is an analytical process for the separation of ions based on charge affinity. IC can be used for a variety of different kinds of charged analytes from single elements to large proteins. In order to ensure accurate analysis, quality standards which are traceable and stable are necessary. Spex CertiPrep®IC standards are of the highest quality available for the analytical laboratory.
All eluents are at 100-fold concentration and ready for dilution, as needed, with filtered ASTM Type I Water.
Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.
Can’t find the standards you are looking for? Spex CertiPrep®can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.
Call For Price
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Concentration Volume (mL) Description
Availability Pricing
COMPARE
0.5 M 125 Sodium Carbonate (Na2CO3) Eluent Concentrate, 0.5 <i>M</i> in H2O; 125 mL
Call For Price
COMPARE
0.18 M 125 Sodium Carbonate (Na2CO3) Eluent Concentrate, 0.18 <i>M</i> in H2O; 125 mL