Spex Purgeable Gases Mixes B
Analytical standards for drinking water, wastewater, and solid waste
  • Accredited by A2LA to ISO/IEC 17025:2017 and ISO 17034:2016
  • Manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
  • For US EPA Methods 502.1, 502.2, 601, 624, 8021, 8260 and CLP Series Methods
US EPA Method 502.1 is a general analytical method for the monitoring of halogenated, portable VOCs in drinking water and raw water sources by GC with a Hall Detector.
US EPA Method 502.2 is a general analytical method for the monitoring of halogenated, purgeable VOCs in drinking water and raw water sources by GC megabore capillary column techniques and Hall and Photoionization Detectors (PID).
US EPA Method 601 is a general analytical method for the monitoring of halogenated VOCs in municipal and industrial wastewater by GC with a Hall Detector.
US EPA Method 624 is a general analytical method for the monitoring of VOCs in municipal and industrial wastewater by GC/MS.
US EPA Method 8021 is a general analytical method for the monitoring of VOCs by GC megabore capillary column techniques and Hall and Photoionization Detectors (PID).
US EPA Method 8260 is a general analytical method for the monitoring of VOCs by GC/MS.
CLP Series Methods: The US EPA retains analytical services through the Contract Laboratory Program (CLP). The CLP follows detailed SOPs derived from EPA methods and SW-846. The CLP Volatile Organic Analysis Method is an analytical method for the analysis of aqueous or solid samples for VOCs by GC/MS.
Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.
Can’t find the standards you are looking for? Spex CertiPrep® can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.
Call For Price
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
CAS Number Concentration Description
Availability Pricing
COMPARE
74-83-9, 75-00-3, 74-87-3, 75-71-8, 75-69-4, 75-01-4 200 µg/mL (200 ppm) Purgeable Gases Mix B, 200 µg/mL in Methanol-P&T; 1 mL
Call For Price
COMPARE
74-83-9, 75-00-3, 74-87-3, 75-71-8, 75-69-4, 75-01-4 2,000 µg/mL (2,000 ppm) Purgeable Gases Mix B (High Level), 2,000 µg/mL in Methanol-P&T; 1mL