Spex Polynuclear Aromatic Hydrocarbon (PAH) Mix, 2,000 µg/mL in Methylene Chloride:Benzene (50:50)

Analytical standard for GC/MS
  • Accredited by A2LA to ISO/IEC 17025:2017 and ISO 17034:2016
  • Manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
All ampulized standards are supplied with a prelabeled transfer/storage vial. We want to make the transfer of ampulized standards as easy as possible. The label includes the catalog number, lot number, product description and expiration date. Custom mixes are also available.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Can’t find the standards you are looking for? Spex CertiPrep® can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.
Call For Price

3 variations of this product are available.

See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Concentration Packaging Volume (mL)
Availability Pricing
COMPARE
2,000 µg/mL (2,000 ppm) 1 x 1.8 mL 1.8
Call For Price
COMPARE
100 µg/mL (100 ppm) 4 x 1 mL 1
Call For Price
COMPARE
2,000 µg/mL (2,000 ppm) 3 x 1.8 mL 1.8
Call For Price
Showing 1- 3 of 3

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At+44 (0)1480 272279
Live chat