Spex Oxalate (C2O4)-2 Ion Anion Standards, 1,000 µg/mL in H2O
Single-element anion standards for ion chromatography
  • Ideal for calibrating and performing QA/QC checks on ion chromatography systems
  • Manufactured under ISO 9001, ISO/IEC 17025 and ISO 17034 compliance
  • Traceable standards ensure accurate analysis
Ion chromatography (IC) is an analytical process for the separation of ions based on charge affinity. IC can be used for a variety of different kinds of charged analytes from single elements to large proteins. In order to ensure accurate analysis, quality standards which are traceable and stable are necessary. SPEX CertiPrep offers the highest quality IC standards available for the analytical laboratory.
Can’t find the standards you are looking for? Spex CertiPrep®can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.
Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.
Call For Price
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Concentration Volume (mL) Description
Availability Pricing
COMPARE
1,000 µg/mL (1,000 ppm) 125 Oxalate (C2O4)-2 Single-Element Ion Anion Standard, 1,000 µg/mL in H2O; 125 mL
Call For Price
COMPARE
1,000 µg/mL (1,000 ppm) 500 Oxalate (C2O4)-2 Single-Element Ion Anion Standard, 1,000 µg/mL in H2O; 500 mL