Spex Multi-Element Organometallic Oil Standards, 5 Elements

Organometallic oil standards for the determination of metals in oils and lubricants
  • Manufactured under an internationally accredited ISO 9001 quality system and compliant with ISO/IEC 17025
  • Guaranteed stable and accurate
  • Clear, transparent mix
The determination of wear metals in engine oils and other lubricants can be applied to machines such as automobiles, aircraft, heavy equipment, trucks, locomotives, military vehicles, etc. The examples are endless. By tracking metals suspended in the used oil, engineers, designers and mechanics can determine the breakdown of specific engine parts. Specific elements present in used oils have been found to be directly related to specific engine problems. Engine failures, as well as expensive repairs, can be avoided if engine oils are analyzed, providing a periodic trend to predict maintenance or replacement.

Applications include:
• Wear metals
• Crude oils
• Additive metals
• Environmental monitoring
• Petrochemical testing
• Pharmaceuticals
• Food processing
• Sulfur in diesel fuel

Can’t find the standards you are looking for? Spex CertiPrep®can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.
Call For Price

5 variations of this product are available.

See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Concentration Weight (g) Description
Availability Pricing
COMPARE
900 µg/g (900 ppm) 100 Organometallic Oil Standard, 5 Elements, 900 µg/g; 100 g
Call For Price
COMPARE
1,000 µg/g (1,000 ppm) 50 Organometallic Oil Standard, 5 Elements, 1,000 µg/g; 50 g
Call For Price
COMPARE
1,000 µg/g (1,000 ppm) 100 Organometallic Oil Standard, 5 Elements, 1,000 µg/g; 100 g
Call For Price
COMPARE
5,000 µg/g (5,000 ppm) 50 Organometallic Oil Standard, 5 Elements, 5,000 µg/g; 50 g
Call For Price
COMPARE
5,000 µg/g (5,000 ppm) 100 Organometallic Oil Standard, 5 Elements, 5,000 µg/g; 100 g
Call For Price
Showing 1- 5 of 5

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At+44 (0)1480 272279
Live chat