Spex Fluoride (F)- Single-Element Ion Anion Standard
Fluoride single-element ion anion standards are of the highest quality
  • Traceable standards
  • Perfect for the analytical laboratory
Ion chromatography (IC) is an analytical process for the separation of ions based on charge affinity. IC can be used for a variety of different kinds of charged analytes from single elements to large proteins. To ensure accurate analysis, quality standards which are traceable and stable are necessary. Spex CertiPrep®offers the highest quality IC standards available for the analytical laboratory.
Can’t find the standards you are looking for? Spex CertiPrep®can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.
Call For Price
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Concentration Element/Ion Volume (mL)
Availability Pricing
COMPARE
100 µg/mL (100 ppm) Fluoride 125
Call For Price
COMPARE
100 µg/mL (100 ppm) Fluoride 500