Spex Claritas PPT® Grade Isotope Standards for ICP-MS

Isotope standards for ICP-MS analysis
  • For isotope dilution analysis and internal standards
The internal standard element must have similar characteristics to the tested/measured element(s) and not be present in the sample. Using isotope modification standards, the chemist can use less internal standard and have a higher intensity reading while avoiding interferences.

Can’t find the standards you are looking for? Spex CertiPrep®can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.
Call For Price

5 variations of this product are available.

See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Element/Ion Volume (mL) Description
Availability Pricing
COMPARE
Boron 11 (B) 125 Claritas PPT® Grade Boron 11, 10 µg/mL in H2O; 125 mL
Call For Price
COMPARE
Copper 65 (Cu) 125 Claritas PPT® Grade Copper 65, 10 µg/mL in 2% HNO3; 125 mL
Call For Price
COMPARE
Lead 206 (Pb) 125 Claritas PPT® Grade Lead 206, 10 µg/mL in 2% HNO3; 125 mL
Call For Price
COMPARE
Lead 207 (Pb) 125 Claritas PPT® Grade Lead 207, 10 µg/mL in 2% HNO3; 125 mL
Call For Price
COMPARE
Zinc 68 (Zn) 125 Claritas PPT® Grade Zinc 68, 10 µg/mL in 2% HNO3; 125 mL
Call For Price
Showing 1- 5 of 5

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At+44 (0)1480 272279
Live chat