Spex Claritas PPT® Grade ICP-MS Multi-Element Standards without Li+6
Multi-element standards for ICP and ICP-MS instrumentation
  • Calibrate ICP-MS instrumentation prior to analysis
  • Dilute to the concentration appropriate for the instrument
  • Manufactured under ISO 9001, ISO/IEC 17025 and ISO 17034 compliance
Spex CertiPrep® Claritas PPT® standards are a class of Inorganic Certified Reference Standards designed specifically for today's new generation of trace ICP and ICP-MS instrumentation. Based on extensive development, our chemists have formulated this line of high-purity standards for user convenience and stability.
Our Claritas PPT® selection of standards includes a complete series of multi-element solutions, many designed for use with US EPA Methods. These solutions are made with the highest purity materials available and are tested on our state-of-the-art ICP-MS. Spex CertiPrep® Certified Reference Materials (CRMs) are manufactured under a quality system complying with the requirements of ISO 9001, ISO/IEC 17025 and ISO 17034.
Call For Price
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Element/Ion Matrix Volume (mL)
Availability Pricing
COMPARE
Bismuth, Germanium, Indium, Scandium, Terbium, Yttrium 5% HNO3 125
Call For Price
COMPARE
Bismuth, Germanium, Indium, Scandium, Terbium, Yttrium 5% HNO3 500
Call For Price
COMPARE
Bismuth, Holmium, Indium, Scandium, Terbium, Yttrium 2% HNO3 125
Call For Price
COMPARE
Bismuth, Holmium, Rhodium, Scandium, Terbium, Yttrium 2% HNO3 125