Spex Carbon Black Reagents
Highly precise reagents ensure accurate carbon black analysis
  • For ASTM Method D1510
  • Matrix is prepared using double-deionized, ASTM Type I Water to provide optimum shelf life
  • Manufactured under ISO 9001, ISO/IEC 17025 and ISO 17034 compliance
The sodium thiosulfate solutions are prepared for ACS Grade, micro-crystalline Na2S2O3. All solutions are prepared gravimetrically using high-accuracy analytical balances to ensure precise target concentrations. Each batch, thoroughly homogenized using a high-speed industrial mixer, provides reliable results from the first bottle to the last.
Our samples are titrated using an automated titrator. The automated dosing drive uses 10,000 steps over a 20 mL volume, so its dosing increment can be as small as 2 µL. For these applications, we are using a minimum dose of 10 µL for the sodium thiosulfate endpoint and 4 µL for the iodine endpoint. These settings achieve the extremely precise measurements for each titration while also staying within the parameters of the dosing unit.
The sodium thiosulfate is run against a 0.1 N potassium dichromate solution. The exact normality of this solution is calculated by comparing it to NIST potassium dichromate. A set of 6 samples are run that must all be within the nominal value of 0.0394 N +/- 0.00008 N. The certified sodium thiosulfate is then used to titrate iodine. A set of 3 samples are run that must all be within the nominal value of 0.0473 N +/- 0.00003 N. Before releasing either of these reagents for packaging, we run QC checks with a previous lot to ensure accuracy over time.
Iodine and sodium thiosulfate solutions may undergo changes in normalities during storage over time even in sealed containers. Once a container is opened, both solutions may change rapidly due to evaporation or oxidation reactions. Open containers exposed to air may incur bacterial growth over time and must be checked before use. ASTM Method D1510 requires that the blanks are checked daily.
Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.
Can’t find the standards you are looking for? Spex CertiPrep®can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.
Call For Price
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Normality Volume (mL) Description
Availability Pricing
COMPARE
0.0394 3785 Sodium Thiosulfate, 0.0394 N in H2O/0.5% Amyl Alcohol; 1 Gallon