Spex Assurance® Grade Quality Control Standard 22 for AA & ICP
Multi-element quality control standards for AA and ICP instrumentation
  • Accredited by A2LA to ISO/IEC 17025/2017 and ISO 17034:2016
  • Certified to DQS to ISO 9001:2015
Quality Control Standards are used to check the standard curve, the procedure for inter-element correction and other spectral interferences. These standards are carried through the entire analytical operation of the method. If the determined concentration is not within ±5% of 1 µg/mL, the laboratory performance is unacceptable. The source of the problem should be identified and corrected before continuing the analysis.
Can’t find the standards you are looking for? Spex CertiPrep®can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.
Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.
Call For Price
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Element/Ion Volume (mL) Description
Availability Pricing
COMPARE
Multiple 125 Assurance® Grade Quality Control Standard 22 for AA & ICP in 5% HNO3/tr. Tartaric Acid/tr. HF; 125 mL
Call For Price
COMPARE
Multiple 250 Assurance® Grade Quality Control Standard 22 for AA & ICP in 5% HNO3/tr. Tartaric Acid/tr. HF; 250 mL
Call For Price
COMPARE
Multiple 500 Assurance® Grade Quality Control Standard 22 for AA & ICP in 5% HNO3/tr. Tartaric Acid/tr. HF; 500 mL