Spex Assurance® Grade Mixed Calibration Standards 5A for AA & ICP
Multi-element calibration standards for AA & ICP instrumentation
  • Manufactured under ISO 9001, ISO/IEC 17025 and ISO 17034 compliance
  • For use in US EPA Method 200.7 (Revision 4.4) and SW-846, Method 6010 (Third Edition)
AA & ICP calibration standards are provided for routine instrument calibration. The concentrations and matrices have been selected for convenience of use and stability.
Can’t find the standards you are looking for? Spex CertiPrep®can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.
Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.
Call For Price
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Element/Ion Volume (mL) Description
Availability Pricing
COMPARE
Beryllium, Iron, Lead, Magnesium, Nickel, and Thallium 125 Assurance® Grade Mixed Calibration Standard 5A for AA & ICP in 5% HNO3; 125 mL
Call For Price
COMPARE
Beryllium, Iron, Lead, Magnesium, Nickel, and Thallium 500 Assurance® Grade Mixed Calibration Standard 5A for AA & ICP in 5% HNO3; 500 mL