Spex Assurance® Grade Instrument Check Standard 10 for AA & ICP
Multi-element instument check standards for AA and ICP instrumentation
  • Accredited by A2LA to ISO/IEC 17025/2017 and ISO 17034:2016
  • Certified to DQS to ISO 9001:2015
  • Calibrate and verify wavelength accuracy and stability in sequential and simultaneous ICP units
Each Instrument Check Standard is designed to give the user wavelength ranges from 160 to 790 nm. Every ICP manufacturer has a specific group of elements at varying concentrations to determine instrument accuracy and reliability. Some have special calibration programs incorporated into their software; others give you information in their manuals.
Can’t find the standards you are looking for? Spex CertiPrep®can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.
Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.
Call For Price
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Element/Ion Volume (mL) Description
Availability Pricing
COMPARE
Multiple 125 Assurance® Grade Instrument Check Standard 10 for AA & ICP in 2% HNO3; 125 mL
Call For Price
COMPARE
Multiple 500 Assurance® Grade Instrument Check Standard 10 for AA & ICP in 2% HNO3; 500 mL