Spex Assurance® Grade Calibration and Matrix Blanks
Establish your baseline for AA and ICP analysis
  • Can be run directly as a blank to establish your baseline
  • May be used to dilute your multi-element standards
Assurance® grade certified calibration and matrix blanks are designed for atomic absorption (AA) and inductively coupled plasma (ICP) analysis. Blanks available include DI water, hydrochloric acid, or nitric acid.
Can’t find the standards you are looking for? Spex CertiPrep®can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.
Call For Price
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Matrix Volume (mL) Description
Availability Pricing
COMPARE
5% HNO3 500 Nitric Acid Blank for AA and ICP in 5% HNO3, 500 mL
Call For Price
COMPARE
5% HCl 500 Hydrochloric Acid Blank for AA and ICP in 5% HCl, 500 mL
Call For Price
COMPARE
H2O 500 DI Water Blank for AA and ICP in H2O, 500 mL
Call For Price
COMPARE
H2O 1 DI Water Blank for AA and ICP in H2O, 1 L
Call For Price
COMPARE
H2O 2 DI Water Blank for AA and ICP in H2O, 2 L
Call For Price
COMPARE
H2O 4 DI Water Blank for AA and ICP in H2O, 4 L