Shimpo Portable LED Stroboscopes with Trigger Light Activation
A quick trigger activates 21 super bright CREE high powered LED lamps
  • Quick and easy flash-control adjustment
  • Measure 30 to 120,000 flashes per minute (fpm) or 0.5 to 2000 Hz
  • Ergonomic large handle grip with trigger style activation
  • Backlit LCD display
  • Fast charge rechargeable battery
  • Accepts external signal to control flash rate
Use these stroboscopes for predictive and preventative maintenance applications such as measuring the speed and frequency of motors, shafts, fans, and much more from a safe distance. Lightweight, portable stroboscopes feature 21 super bright LED lamps that provide superior service life compared to xenon-lit stroboscopes.
Simply press the power trigger button to activate or deactivate the light arrays. Use the “X” and “÷” buttons to quickly multiply or divide the flash rate by 2; fine tune flash rate in single-digit increments using “+” or “–” buttons. Ergonomically designed large handle grip minimizes user fatigue even during extended use. The external trigger mode allows the flash rate to be controlled by an external signal. The flash rate automatically increases or decreases depending on the received signal.
£389.93 - £501.93GBP / Each
£467.92 - £602.32GBP (incl VAT)
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Flash Duration Max Flash Range (fpm) Description
Availability Pricing
Shimpo ST-4000 Portable LED Stroboscope with Trigger Activation 2530499 Mfr # ST-4000
COMPARE
0.1 to 2.5° = 1 to 100 on display 120000 Digital LED Stroboscope with Trigger Activation
£389.93GBP / Each
£467.92GBP (incl VAT)
Shimpo ST-5100 Digital LED Stroboscope with Trigger Activation and Auto-Speed Detection 2530517 Mfr # ST-5100
COMPARE
0.1 to 2.5° = 1 to 100 on display 120000 Digital LED Stroboscope with Trigger Activation and Auto-Speed Detection
£501.93GBP / Each
£602.32GBP (incl VAT)