SE International Radiation Alert™ Monitor 200 Radiation Meter
Lightweight and rugged—ideal for the lab or field
  • Sensitive to alpha, beta, gamma, and x-rays to keep you safe
  • Backlit display allows you to see readings, even in low-light conditions
  • Red light flashes, and alert beeps with each count detected
  • Adjustable alert level and timer
  • Download data, set alarms, and calibrate with free software
Rugged enough to survey the field, with all the features of a lab unit. The digital display features large digits and selectable backlight—read under any conditions. A red LED flashes and a 70 dB alert beeps with each count detected (may be muted). Icons also make mode and conditions easy to understand.||Save individual readings or timed captures to the internal memory and export to computer through USB with the free software. The software also allows you to set alarm levels and calibrate the meter. Both a cable and software are included.
£682.79GBP / Each
£819.35GBP (incl VAT)
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Sensor Description
Availability Pricing
Radiation Alert Monitor 200 Digital Radiation Meter and Data Logger 8191024 Mfr # MONITOR 200
COMPARE
Internal Digital Radiation Meter and Data Logger
£682.79GBP / Each
£819.35GBP (incl VAT)