Scienceware Vakuwash® Cuvette Washer
Protect your sensitive optical surfaces
  • Constructed of durable polyethylene
This washer gently and thoroughly cleans cuvettes without the risk of scratching or chipping. Durable polyethylene housing resists most detergents and solvents. Simply connect washer to your vacuum pump or aspirator, place mouth of cell onto rubber cushion to induce suction, and pour water or cleaning fluid into funnel. The fluid will spray forcibly into the cell and drain into the filter flask - eliminating cross contamination and protecting you from hazardous fluids. Air dry or auto-dry with continued suction.
£193.52GBP / Each
£232.22GBP (incl VAT)
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Max Capacity (mL) Max Capacity (MicroL) Pathlength (mm)
Availability Pricing
Scienceware 38960-0000 PE Cuvette/Cell Washer with 500 mL Receiver Flask 8330000 Mfr # 38960-0000
COMPARE
£193.52GBP / Each
£232.22GBP (incl VAT)