Savillex Lab Bottle with Closure, FEP
Ultra-pure FEP bottles with exceptional clarity and wide temperature range for critical applications
  • FEP material provides excellent chemical resistance
  • Withstands temperatures as low as –200°C (–328°F)
  • Stretch blow molded for smooth surface finish
  • Superior optical clarity for easy content visibility
  • Leakproof FEP screw closure
  • Manufactured and sealed in bags inside a cleanroom
Savillex FEP lab bottles deliver unmatched chemical purity, optical clarity, and a wide temperature range for demanding applications. Manufactured using a unique stretch blow molding process, these high-performance bottles offer lower trace metal content, smoother surface finish, easier pouring, and superior sealing. The crystal-clear FEP material provides excellent visibility of bottle contents while maintaining the chemical resistance and purity required for critical lab work. The linerless FEP screw closures provide a secure, leak-free seal. Working temperature range is from -200 to 200°C (-328 to 392°F).
£67.62 - £359.23GBP / Each
£81.14 - £431.08GBP (incl VAT)
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Cap / Stopper Size Material
Availability Pricing
Savillex Lab Bottle with GL45 Closure, FEP, 100 mL  0605208 Mfr # 150-02-0100
COMPARE
100 GL45 Fluorinated Ethylene Propylene (FEP)
£67.62GBP / Each
£81.14GBP (incl VAT)
Savillex Lab Bottle with GL45 Closure, FEP, 250 mL  0605209 Mfr # 150-02-0250
COMPARE
250 GL45 Fluorinated Ethylene Propylene (FEP)
£102.05GBP / Each
£122.46GBP (incl VAT)
Savillex Lab Bottle with GL45 Closure, FEP, 500 mL 0605210 Mfr # 150-02-0500
COMPARE
500 GL45 Fluorinated Ethylene Propylene (FEP)
£136.48GBP / Each
£163.78GBP (incl VAT)
Savillex Lab Bottle with GL45 Closure, FEP, 1000 mL 0605211 Mfr # 150-02-1000
COMPARE
1000 GL45 Fluorinated Ethylene Propylene (FEP)
£205.13GBP / Each
£246.16GBP (incl VAT)
Savillex Bottle with GL45 Closure, FEP, 2000 mL 0605212 Mfr # 150-02-2000
COMPARE
2000 GL45 Fluorinated Ethylene Propylene (FEP)
£359.23GBP / Each
£431.08GBP (incl VAT)