Sash Stop for Labconco Fume Hoods
Restricts how far a vertical-rising sash may be opened
  • Easy to install on fixture panel
Sash stop restricts the sash opening height of your fume hood. With the sash kept in a lowered position, the exhaust volume demand on the blower is reduced. Release the sash stop to open the sash completely for easier hood loading and cleaning. Sash stop kit can be used on most Labconco fume hoods made since 2012.
£65.84GBP / Each
£79.01GBP (incl VAT)
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Exterior Height (in) Exterior Width (in) Power (VAC)
Availability Pricing
Labconco 9410300 Sash Stop for Protector Premier Hoods 0907084 Mfr # 9410300
COMPARE
£65.84GBP / Each
£79.01GBP (incl VAT)