Sartorius® Quintix Pro Precision Top Loading Balances
Designed for precision and usability - suitable for wide range of laboratory applications
  • Easy access to menu option with 7” high resolution touch display
  • Automatic internal adjustment (isoCAL) delivers accurate results
  • Stand-by or switch-off modes saves energy
  • Chemically-resistant materials offer easy cleaning and maintenance
  • Automatic dim function reduces display brightness, saves power
  • Built-in interactive manual allows quick access to information
The Quintix Pro Precision toploading balances are designed for usability and precision, making them ideal for a variety of laboratory applications. They feature a 7-inch high-resolution touch display with a user-friendly interface, ensuring easy navigation. Key functionalities include an internal motorized leveling system (IMO & IMU models) and automatic internal adjustment (isoCAL) for optimal accuracy. The LevelControl function monitors balance stability in real-time, while the semi-automatic filter adaptation (AFA) allows customization for specific tasks.
Data transfer is seamless, with multiple connection options including three USB Type C interfaces, RS-232, and ethernet. The balances support GLP/GMP-compliant printouts and facilitate data transfer to USB or databases.
These balances are versatile enough for cleanrooms (grade A and B) and specialized environments like vacuum, argon, and nitrogen settings. They come with an extended language package (15 languages) and 15 integrated application programs for various weighing tasks. Security features include customizable access levels with passcode protection.
Additional features, such as stand-by mode, automatic dimming, and a user-defined start timer, promote energy efficiency. Made with recycled materials and eco-friendly packaging, the Quintix Pro Precision balances prioritize sustainability without compromising on performance, making them an excellent choice for any laboratory setting.
£2,441.19 - £5,967.28GBP / Each
£2,929.43 - £7,160.74GBP (incl VAT)
30 variations of this product are available.
See All Product Options Below
30 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (g) Readability (mg) Description
Availability Pricing
Sartorius Quintix Pro QTX1503IRU-1S Precision Toploading, 1500g x 1.0mg, Internal Calibration and Guided Leveling 1197702 Mfr # QTX1503IRU-1S
COMPARE
1500 1 Precision Toploading, 1500g x 1.0mg, with Internal Calibratio and Guided Leveling
£5,243.97GBP / Each
£6,292.76GBP (incl VAT)
Sartorius Quintix Pro QTX1203IRU-1S Precision Toploading, 1200g x 1.0mg, Internal Calibration and Guided Leveling 1197703 Mfr # QTX1203IRU-1S
COMPARE
1200 1 Precision Toploadin, 1200g x 1.0mg, with Internal Calibration and Guided Leveling
£4,972.72GBP / Each
£5,967.26GBP (incl VAT)
Sartorius Quintix Pro QTX623IRU-1S Precision Toploading  Balance, 620g x 1.0mg, Internal Calibration and Guided Leveling 1197704 Mfr # QTX623IRU-1S
COMPARE
620 1 Precision Toploading Balance, 620g x 1.0mg, with Internal Calibration and Guided Leveling
£4,068.59GBP / Each
£4,882.31GBP (incl VAT)
Sartorius Quintix Pro QTX423IRU-1S Precision Toploading  Balance, 420g x 1.0mg, Internal Calibration and Guided Leveling 1197705 Mfr # QTX423IRU-1S
COMPARE
420 1 Precision Toploading Balance, 420g x 1.0mg, with Internal Calibration and Guided Leveling
£3,616.53GBP / Each
£4,339.84GBP (incl VAT)
Sartorius Quintix Pro QTX323IRU-1S Precision Toploading  Balance, 320g x 1.0mg, Internal Calibration and Guided Leveling 1197706 Mfr # QTX323IRU-1S
COMPARE
320 1 Precision Toploading Balance, 320g x 1.0mg, with Internal Calibration and Guided Leveling
£3,164.46GBP / Each
£3,797.35GBP (incl VAT)
Sartorius Quintix Pro QTX6202IRO-1S Precision Toploading Balance, 6200g x 10mg, Internal Calibration and Guided Leveling (no draft shield) 1197707 Mfr # QTX6202IRO-1S
COMPARE
6200 10 Precision Toploading Balance, 6200g x 10mg, with Internal Calibration and Guided Leveling (no draft shield)
£4,068.59GBP / Each
£4,882.31GBP (incl VAT)
Sartorius Quintix Pro QTX4202IRO-1S Precision Toploading Balance, 4200g x 10mg, Internal Calibration and Guided Leveling (no draft shield) 1197708 Mfr # QTX4202IRO-1S
COMPARE
4200 10 Precision Toploading Balance, 4200g x 10mg, with Internal Calibration and Guided Leveling (no draft shield)
£3,616.53GBP / Each
£4,339.84GBP (incl VAT)
Sartorius Quintix Pro QTX3202IRO-1S Precision Toploading Balance, 3200g x 10mg, Internal Calibration and Guided Leveling (no draft shield) 1197709 Mfr # QTX3202IRO-1S
COMPARE
3200 10 Precision Toploading Balance, 3200g x 10mg, with Internal Calibration and Guided Leveling (no draft shield)
£3,164.46GBP / Each
£3,797.35GBP (incl VAT)
Sartorius Quintix Pro QTX2202IRO-1S Precision Toploading Balance, 2200g x 10mg, Internal Calibration and Guided Leveling (no draft shield) 1197710 Mfr # QTX2202IRO-1S
COMPARE
2200 10 Precision Toploading Balance, 2200g x 10mg, with Internal Calibration and Guided Leveling (no draft shield)
£2,712.40GBP / Each
£3,254.88GBP (incl VAT)
Sartorius Quintix Pro QTX1202IRO-1S Precision Toploading Balance, 1200g x 10mg, Internal Calibration and Guided Leveling (no draft shield) 1197711 Mfr # QTX1202IRO-1S
COMPARE
1200 10 Precision Toploading Balance, 1200g x 10mg, with Internal Calibration and Guided Leveling (no draft shield)
£2,441.19GBP / Each
£2,929.43GBP (incl VAT)
Sartorius Quintix Pro QTX12201IRO-1S Precision Toploading Balance, 12,200g x 0.1g, Internal Calibration and Guided Leveling (no draft shield) 1197712 Mfr # QTX12201IRO-1S
COMPARE
12200 100
£4,068.59GBP / Each
£4,882.31GBP (incl VAT)
Sartorius Quintix Pro QTX10201IRO-1S Precision Toploading Balance, 10,200g x 0.1g, Internal Calibration and Guided Leveling (no draft shield) 1197713 Mfr # QTX10201IRO-1S
COMPARE
10200 100 Precision Toploading Balance, 10,200g x 0.1g, with Internal Calibration and Guided Leveling (no draft shield)
£3,345.32GBP / Each
£4,014.38GBP (incl VAT)
Sartorius Quintix Pro QTX8201IRO-1S Precision Toploading Balance, 8,200g x 0.1g, Internal Calibration and Guided Leveling (no draft shield) 1197714 Mfr # QTX8201IRO-1S
COMPARE
8200 100 Precision Toploading Balance, 8,200g x 0.1g, with Internal Calibration and Guided Leveling (no draft shield)
£3,164.46GBP / Each
£3,797.35GBP (incl VAT)
Sartorius Quintix Pro QTX6201IRO-1S Precision Toploading Balance, 6,200g x 0.1g, Internal Calibration and Guided Leveling (no draft shield) 1197715 Mfr # QTX6201IRO-1S
COMPARE
6200 100 Precision Toploading Balance, 6,200g x 0.1g, with Internal Calibration and Guided Leveling (no draft shield)
£2,712.40GBP / Each
£3,254.88GBP (incl VAT)
Sartorius Quintix Pro QTX3201IRO-1S Precision Toploading Balance, 3,200g x 0.1g, Internal Calibration and Guided Leveling (no draft shield) 1197716 Mfr # QTX3201IRO-1S
COMPARE
3200 100 Precision Toploading Balance, 3,200g x 0.1g, with Internal Calibration and Guided Leveling (no draft shield)
£2,441.19GBP / Each
£2,929.43GBP (incl VAT)
Sartorius Quintix Pro QTX1503IMU-1S Precision Toploading  Balance, 1500g x 1.0mg, Internal Calibration and Motorized Leveling 1197720 Mfr # QTX1503IMU-1S
COMPARE
1500 1 Precision Toploading Balance, 1500g x 1.0mg, with Internal Calibration and Motorized Leveling
£5,967.28GBP / Each
£7,160.74GBP (incl VAT)
Sartorius Quintix Pro QTX1203IMU-1S Precision Toploading  Balance, 1200g x 1.0mg, Internal Calibration and Motorized Leveling 1197721 Mfr # QTX1203IMU-1S
COMPARE
1200 1 Precision Toploading Balance, 1200g x 1.0mg, with Internal Calibration and Motorized Leveling
£5,696.03GBP / Each
£6,835.24GBP (incl VAT)
Sartorius Quintix Pro QTX623IMU-1S Precision Toploading  Balance, 620g x 1.0mg, Internal Calibration and Motorized Leveling 1197722 Mfr # QTX623IMU-1S
COMPARE
620 1 Precision Toploading Balance, 620g x 1.0mg, with Internal Calibration and Motorized Leveling
£4,791.90GBP / Each
£5,750.28GBP (incl VAT)
Sartorius Quintix Pro QTX423IMU-1S Precision Toploading  Balance, 420g x 1.0mg, Internal Calibration and Motorized Leveling 1197723 Mfr # QTX423IMU-1S
COMPARE
420 1 Precision Toploading Balance, 420g x 1.0mg, with Internal Calibration and Motorized Leveling
£4,339.84GBP / Each
£5,207.81GBP (incl VAT)
Sartorius Quintix Pro QTX323IMU-1S Precision Toploading  Balance, 320g x 1.0mg, Internal Calibration, and Motorized Leveling 1197724 Mfr # QTX323IMU-1S
COMPARE
320 1 Precision Toploading Balance, 320g x 1.0mg, with Internal Calibration, and Motorized Leveling
£3,887.77GBP / Each
£4,665.32GBP (incl VAT)
Showing 1 - 20 of 30