Sani-Tech® STHT®-R Braid-Reinforced Platinum-Cured Silicone Tubing
Excellent for high-purity fluid transfers at elevated pressures
  • Braid reinforced yet remains ultra flexible
  • Resistant to temperature extremes, radiation, and chemicals
  • Platinum cured silicone—shows exceptional biocompatibility
  • Meets the requirements of USP <88> Class VI, and/or USP <87>, and/or ISO 10993-5
  • Produced and packed in an ISO Class 7 certified cleanroom
  • Ideal for general-purpose, laboratory, and single-use applications
  • Free technical application support available to assist with tubing selection cleaning and sterilization
Sani-Tech STHT-R silicone tubing is braid reinforced to withstand higher working pressure. Manufactured from Sani-Tech® 65 silicone resin, it exhibits excellent resistance to temperature extremes, radiation, moisture, compression sets, and chemicals. Tubing comes with labels on both the shipping box and bags for easy identification.
£424.89 - £2,595.56GBP / Pkg of 1
£509.87 - £3,114.67GBP (incl VAT)
18 variations of this product are available.
See All Product Options Below
18 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Length (ft) Tubing ID (in) Tubing OD (in)
Availability Pricing
SANI-TECH STHT®-R Reinforced Platinum-Cured Silicone Tubing, 1/8" ID x 23/64" OD; 25 Ft 5020215 Mfr # STHT-R-0125
COMPARE
25 1/8 23/64
£472.00GBP / Pkg of 1
£566.40GBP (incl VAT)
SANI-TECH STHT®-R Reinforced Platinum-Cured Silicone Tubing, 1/8" ID x 23/64" OD; 50 Ft 5020216 Mfr # STHT-R-0125
COMPARE
50 1/8 23/64
£933.34GBP / Pkg of 1
£1,120.01GBP (incl VAT)
SANI-TECH STHT-R Reinforced Platinum-Cured Silicone Tubing, 3/16" ID x 29/64" OD; 25 Ft 5020217 Mfr # F09764
COMPARE
25 3/16 29/64
£424.89GBP / Pkg of 1
£509.87GBP (incl VAT)
SANI-TECH STHT®-R Reinforced Platinum-Cured Silicone Tubing, 3/16" ID x 29/64" OD; 50 Ft 5020218 Mfr # F09764
COMPARE
50 3/16 29/64
£837.34GBP / Pkg of 1
£1,004.81GBP (incl VAT)
SANI-TECH STHT®-R Reinforced Platinum-Cured Silicone Tubing, 1/4" ID x 1/2" OD; 25 Ft 5020219 Mfr # F09766
COMPARE
25 1/4 1/2
£498.67GBP / Pkg of 1
£598.40GBP (incl VAT)
SANI-TECH STHT®-R Reinforced Platinum-Cured Silicone Tubing, 1/4" ID x 1/2" OD; 50 Ft 5020220 Mfr # F09767
COMPARE
50 1/4 1/2
£1,065.75GBP / Pkg of 1
£1,278.90GBP (incl VAT)
SANI-TECH STHT®-R Reinforced Platinum-Cured Silicone Tubing, 3/8" ID x 5/8" OD; 25 Ft 5020221 Mfr # F09785
COMPARE
25 3/8 5/8
£616.89GBP / Pkg of 1
£740.27GBP (incl VAT)
SANI-TECH STHT®-R Reinforced Platinum-Cured Silicone Tubing, 3/8" ID x 5/8" OD; 50 Ft 5020222 Mfr # F09786
COMPARE
50 3/8 5/8
£1,191.12GBP / Pkg of 1
£1,429.34GBP (incl VAT)
SANI-TECH STHT®-R Reinforced Platinum-Cured Silicone Tubing, 1/2" ID x 7/8" OD; 25 Ft 5020223 Mfr # F09788
COMPARE
25 1/2 7/8
£902.23GBP / Pkg of 1
£1,082.68GBP (incl VAT)
SANI-TECH STHT®-R Reinforced Platinum-Cured Silicone Tubing, 1/2" ID x 7/8" OD; 50 Ft 5020224 Mfr # F09671
COMPARE
50 1/2 7/8
£1,782.23GBP / Pkg of 1
£2,138.68GBP (incl VAT)
SANI-TECH STHT®-R Reinforced Platinum-Cured Silicone Tubing, 5/8" ID x 1" OD; 25 Ft 5020225 Mfr # F09791
COMPARE
25 5/8 1
£902.23GBP / Pkg of 1
£1,082.68GBP (incl VAT)
SANI-TECH STHT®-R Reinforced Platinum-Cured Silicone Tubing, 5/8" ID x 1" OD; 50 Ft 5020226 Mfr # F09792
COMPARE
50 5/8 1
£1,786.67GBP / Pkg of 1
£2,144.00GBP (incl VAT)
SANI-TECH STHT®-R Reinforced Platinum-Cured Silicone Tubing, 3/4" ID x 1-1/8" OD; 25 Ft 5020227 Mfr # F09793
COMPARE
25 3/4 1 1/8
£1,057.78GBP / Pkg of 1
£1,269.34GBP (incl VAT)
SANI-TECH STHT®-R Reinforced Platinum-Cured Silicone Tubing, 3/4" ID x 1-1/8" OD; 50 Ft 5020228 Mfr # F09794
COMPARE
50 3/4 1 1/8
£2,115.56GBP / Pkg of 1
£2,538.67GBP (incl VAT)
SANI-TECH STHT®-R Reinforced Platinum-Cured Silicone Tubing, 7/8" ID x 1-1/4" OD; 25 Ft 5020229 Mfr # F09795
COMPARE
25 7/8 1 1/4
£1,115.56GBP / Pkg of 1
£1,338.67GBP (incl VAT)
SANI-TECH STHT®-R Reinforced Platinum-Cured Silicone Tubing, 7/8" ID x 1-1/4" OD; 50 Ft 5020230 Mfr # F09796
COMPARE
50 7/8 1 1/4
£2,226.67GBP / Pkg of 1
£2,672.00GBP (incl VAT)
SANI-TECH STHT®-R Reinforced Platinum-Cured Silicone Tubing, 1" ID x 1-3/8" OD; 25 Ft 5020231 Mfr # F09797
COMPARE
25 1 1 3/8
£1,280.00GBP / Pkg of 1
£1,536.00GBP (incl VAT)
SANI-TECH STHT-R Reinforced Platinum-Cured Silicone Tubing, 1" ID x 1-3/8" OD; 50 Ft 5020232 Mfr # F09798
COMPARE
50 1 1 3/8
£2,595.56GBP / Pkg of 1
£3,114.67GBP (incl VAT)
Showing 1- 18 of 18