Sani-Tech® STHT®-C Platinum-Cured Pharmaceutical Silicone Tubing
Sani-Tech STHT-C silicone tubing features ultra-pure and ultra-smooth inner bore, perfect for biopharmaceutical applications
  • Ultra-smooth inner surface minimizes particle entrapment
  • Durable tubing withstands repeated autoclaving
  • Platinum cured silicone—shows exceptional biocompatibility
  • Low extractables—perfect for maintaining fluid purity
  • Meets the requirements of USP <88> Class VI, and/or USP <87>, and/or ISO 10993-5
  • Produced and double-bagged in an ISO Class 7 certified cleanroom
  • Ideal for general-purpose, laboratory, and single-use applications
  • Free technical application support available to assist with tubing selection cleaning and sterilization
Sani-Tech STHT-C tubing, manufactured from the finest grade of silicone material, features ultra-pure, ultra-smooth inner surface—perfect transferring biological fluids. The smooth inner surface ensures minimal particle entrapment and microscopic build-up during fluid transfer while low extractables help maintain fluid integrity. This durable tubing withstands repeated autoclaving to help maintain the integrity of your sample.
£201.78GBP / Pkg of 1
£242.14GBP (incl VAT)
37 variations of this product are available.
See All Product Options Below
37 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Tubing ID (in) Tubing OD (in) Wall Thickness (in)
Availability Pricing
Sani-Tech® STHT®-C Platinum-Cured Pharmaceutical Silicone Tubing, 0.012" ID x 0.024" OD; 25 Ft 5020130 Mfr # STHT-C-012-0
COMPARE
1/100 1/50 1/200
£53.82GBP / Pkg of 1
£64.58GBP (incl VAT)
Sani-Tech® STHT®-C Platinum-Cured Pharmaceutical Silicone Tubing, 0.02" ID x 0.036" OD; 25 Ft 5020131 Mfr # STHT-C-020-0
COMPARE
1/50 1/28 1/150
£53.82GBP / Pkg of 1
£64.58GBP (incl VAT)
Sani-Tech® STHT®-C Platinum-Cured Pharmaceutical Silicone Tubing, 0.03" ID x 0.066" OD; 25 Ft 5020133 Mfr # STHT-C-030-0
COMPARE
1/32 1/15 1/64
£52.27GBP / Pkg of 1
£62.72GBP (incl VAT)
Sani-Tech® STHT®-C Platinum-Cured Pharmaceutical Silicone Tubing, 0.03" ID x 0.155" OD; 25 Ft 5020134 Mfr # STHT-C-030-2
COMPARE
1/32 5/32 1/16
£69.69GBP / Pkg of 1
£83.63GBP (incl VAT)
Sani-Tech® STHT®-C Platinum-Cured Pharmaceutical Silicone Tubing, 0.04" ID x 0.086" OD; 25 Ft 5020135 Mfr # STHT-C-040-0
COMPARE
1/25 1/12 1/50
£54.14GBP / Pkg of 1
£64.97GBP (incl VAT)
Sani-Tech® STHT®-C Platinum-Cured Pharmaceutical Silicone Tubing, 0.058" ID x 0.076" OD; 25 Ft 5020136 Mfr # STHT-C-058-0
COMPARE
1/16 5/64 1/100
Call For Price
Sani-Tech® STHT®-C Platinum-Cured Pharmaceutical Silicone Tubing, 1/16" ID x 1/8" OD; 25 Ft 5020137 Mfr # STHT-C-062-1
COMPARE
1/16 1/8 1/32
£53.25GBP / Pkg of 1
£63.90GBP (incl VAT)
Sani-Tech® STHT®-C Platinum-Cured Pharmaceutical Silicone Tubing, 1/16" ID x 3/16" OD; 25 Ft 5020138 Mfr # STHT-C-062-2
COMPARE
1/16 3/16 1/16
£76.54GBP / Pkg of 1
£91.85GBP (incl VAT)
Sani-Tech® STHT®-C Platinum-Cured Pharmaceutical Silicone Tubing, 1/16" ID x 1/4" OD; 25 Ft 5020139 Mfr # STHT-C-062-3
COMPARE
1/16 1/4 3/32
£102.67GBP / Pkg of 1
£123.20GBP (incl VAT)
Sani-Tech® STHT®-C Platinum-Cured Pharmaceutical Silicone Tubing, 5/64" ID x 17/64" OD; 25 Ft 5020145 Mfr # STHT-C-078-3
COMPARE
5/64 17/64 3/32
£109.34GBP / Pkg of 1
£131.21GBP (incl VAT)
Sani-Tech® STHT®-C Platinum-Cured Pharmaceutical Silicone Tubing, 3/32" ID x 5/32" OD; 25 Ft 5020148 Mfr # STHT-C-093-1
COMPARE
3/32 5/32 1/32
£64.45GBP / Pkg of 1
£77.34GBP (incl VAT)
Sani-Tech® STHT®-C Platinum-Cured Pharmaceutical Silicone Tubing, 3/32" ID x 7/32" OD; 25 Ft 5020149 Mfr # STHT-C-093-2
COMPARE
3/32 7/32 1/16
£84.98GBP / Pkg of 1
£101.98GBP (incl VAT)
Sani-Tech® STHT®-C Platinum-Cured Pharmaceutical Silicone Tubing, 1" ID x 1-1/8" OD; 25 Ft 5020153 Mfr # STHT-C-1000-2
COMPARE
1 1 1/8 1/16
Call For Price
Sani-Tech® STHT®-C Platinum-Cured Pharmaceutical Silicone Tubing, 1" ID x 1-3/8" OD; 25 Ft 5020156 Mfr # STHT-C-1000-5
COMPARE
1 1 3/8 3/16
£1,035.56GBP / Pkg of 1
£1,242.67GBP (incl VAT)
Sani-Tech® STHT®-C Platinum-Cured Pharmaceutical Silicone Tubing, 1" ID x 1-1/2" OD; 25 Ft 5020157 Mfr # STHT-C-1000-6
COMPARE
1 1 1/2 1/4
£1,435.56GBP / Pkg of 1
£1,722.67GBP (incl VAT)
Sani-Tech® STHT®-C Platinum-Cured Pharmaceutical Silicone Tubing, 1/8" ID x 3/16" OD; 25 Ft 5020158 Mfr # STHT-C-125-1
COMPARE
1/8 3/16 1/32
£67.20GBP / Pkg of 1
£80.64GBP (incl VAT)
Sani-Tech® STHT®-C Platinum-Cured Pharmaceutical Silicone Tubing, 1/8" ID x 1/4" OD; 25 Ft 5020159 Mfr # STHT-C-125-2
COMPARE
1/8 1/4 1/16
£83.12GBP / Pkg of 1
£99.74GBP (incl VAT)
Sani-Tech® STHT®-C Platinum-Cured Pharmaceutical Silicone Tubing, 1/8" ID x 5/16" OD; 25 Ft 5020160 Mfr # STHT-C-125-3
COMPARE
1/8 5/16 3/32
£127.12GBP / Pkg of 1
£152.54GBP (incl VAT)
Sani-Tech® STHT®-C Platinum-Cured Pharmaceutical Silicone Tubing, 1/8" ID x 3/8" OD; 25 Ft 5020161 Mfr # STHT-C-125-4
COMPARE
1/8 3/8 1/8
£169.12GBP / Pkg of 1
£202.94GBP (incl VAT)
Sani-Tech® STHT®-C Platinum-Cured Pharmaceutical Silicone Tubing, 3/16" ID x 1/4" OD; 25 Ft 5020168 Mfr # STHT-C-187-1
COMPARE
3/16 1/4 1/32
Call For Price
Showing 1 - 20 of 37