Saint-Gobain Pure-Fit® SIB® Hose Barb Fittings, Tee Union, Polypropylene
Smooth inner bore ensures seamless fluid transition between tubing and fittings
  • Unique internal design eliminates dead space
  • No areas of entrapment—prohibiting a potential for bacterial growth
  • Use with Pure-Fit and BarbLock® retainers for leak-free connections
  • Easily combine or split fluid flow with a tee union
  • Steriliza by gamma irradiation
  • Animal-derived component free and BPA free
  • Manufactured and packaged in Class 8 cleanroom
  • USP <88> Class VI and USP <87> compliant
  • Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system
  • Free technical application support available to determine which connector type is needed for your application
Eliminate dead space and entrapment, a potential breeding ground for bacterial growth, with smooth inner bore (SIB) fittings. SIB fittings provide a seamless, “zero” transition between tubing and fittings, maintaining complete fluid integrity. Use with Pure-Fit or BarbLock retainer assembly that provides 360° compression to ensure a leak-free connection.
£227.34 - £319.56GBP / Pkg of 25
£272.81 - £383.47GBP (incl VAT)
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
Compareitem specifications
Barb Size / Tubing Id (Port 1) (in)
Availability Pricing
Saint Gobain Pure-Fit® SIB® Hose Barb Fitting, Tee Union, Polypropylene, 1/8"; 25/Pk 5014310 Mfr # PFT125PPAF
COMPARE
1/8
£227.34GBP / Pkg of 25
£272.81GBP (incl VAT)
Saint Gobain Pure-Fit® SIB® Hose Barb Fitting, Tee Union, Polypropylene, 3/16"; 25/Pk 5014317 Mfr # PFT187PPAF
COMPARE
3/16
£230.89GBP / Pkg of 25
£277.07GBP (incl VAT)
Saint Gobain Pure-Fit® SIB® Hose Barb Fitting, Tee Union, Polypropylene, 1/4"; 25/Pk 5014322 Mfr # PFT250PPAF
COMPARE
1/4
£234.67GBP / Pkg of 25
£281.60GBP (incl VAT)
Saint Gobain Pure-Fit® SIB® Hose Barb Fitting, Tee Union, Polypropylene, 5/16; 25/Pk 5014331 Mfr # PFT312PPAF
COMPARE
5/16
£240.89GBP / Pkg of 25
£289.07GBP (incl VAT)
Saint Gobain Pure-Fit® SIB® Hose Barb Fitting, Tee Union, Polypropylene, 3/8"; 25/Pk 5014334 Mfr # PFT375PPAF
COMPARE
3/8
£246.89GBP / Pkg of 25
£296.27GBP (incl VAT)
Saint Gobain Pure-Fit® SIB® Hose Barb Fitting, Tee Union, Polypropylene, 1/2"; 25/Pk 5014342 Mfr # PFT500PPAF
COMPARE
1/2
£260.23GBP / Pkg of 25
£312.28GBP (incl VAT)
Saint Gobain Pure-Fit® SIB® Hose Barb Fitting, Tee Union, Polypropylene, 3/4"; 25/Pk 5014351 Mfr # PFT750PPAF
COMPARE
3/4
£319.56GBP / Pkg of 25
£383.47GBP (incl VAT)
Showing 1- 7 of 7