Roscid Technologies DewTran® Chilled Mirror Hygrometer
A low-cost, precision chilled mirror hygrometer technology that provides quick, accurate, and versatile measurements of dew point, air temperature, humidity, and barometric pressure, all traceable to NIST standards
  • Ensures reliable and accurate measurements
  • Features two SPDT alarms for critical notifications and two configurable 4-20mA outputs
  • Allows for programming and integration with RS-232 connection
  • Equipped with a two-line display for easy reading
  • Capable of logging measurement data for analysis
  • It operates effectively within a wide temperature range from -40°C to 120°C
Discover the DewTran® Series, a patented, low-cost, precision chilled mirror hygrometer. Designed for quick and accurate measurements, it reliably tracks dew point, air temperature, humidity, and barometric pressure. Versatile mounting options include wall, duct, remote, or flow-through applications. Customize your display to show up to three parameters: dew point (DP), air temperature (T), and relative humidity (RH) or barometric pressure (BP), with additional units available. Enjoy stable, accurate, and repeatable measurements that are traceable to NIST, ensuring top-tier reliability and performance.
Call For Price
9 variations of this product are available.
See All Product Options Below
9 PRODUCT OPTIONS
Compareitem specifications
Data Logging Mounting Type Description
Availability Pricing
Roscid Technologies DewTran® Chilled Mirror Hygrometer, Duct Mount 7893049 Mfr # DewTran-R-D
COMPARE
SD 64GB, 1 sample/2 sec, CSV file output Duct Chilled Mirror Hygrometer, Duct Mount
Call For Price
Roscid Technologies DewTran® Chilled Mirror Hygrometer, Wall Mount 7893050 Mfr # DewTran-W
COMPARE
SD 64GB, 1 sample/2 sec, CSV file output Wall Chilled Mirror Hygrometer, Wall Mount
Call For Price
Roscid Technologies DewTran® Chilled Mirror Hygrometer, Wall Mount with Flow-Through Chamber 7893051 Mfr # DewTran-W-FT
COMPARE
SD 64GB, 1 sample/2 sec, CSV file output Wall Chilled Mirror Hygrometer, Wall Mount with Flow-Through Chamber
Call For Price
Roscid Technologies DewTran® Chilled Mirror Hygrometer, Remote Mount with Air Temperature Probe 7893052 Mfr # DewTran-R-T
COMPARE
SD 64GB, 1 sample/2 sec, CSV file output Remote Chilled Mirror Hygrometer, Remote Mount with Air Temperature Probe
Call For Price
Roscid Technologies DewTran® Chilled Mirror Hygrometer, Remote Mount with 15 ft Cable 7893053 Mfr # DewTran-R-15ft
COMPARE
SD 64GB, 1 sample/2 sec, CSV file output Remote Chilled Mirror Hygrometer, Remote Mount with 15 ft Cable
Call For Price
Roscid Technologies DewTran® Chilled Mirror Hygrometer, Remote Mount with Self-Aspirating Fan 7893054 Mfr # DewTran-R-F
COMPARE
SD 64GB, 1 sample/2 sec, CSV file output Remote Chilled Mirror Hygrometer, Remote Mount with Self-Aspirating Fan
Call For Price
Roscid Technologies DewTran® Chilled Mirror Hygrometer, Remote Mount, Tested up to 90°C at 95% RH, with Air Temperature Probe 7893055 Mfr # DewTran-R-T-X
COMPARE
SD 64GB, 1 sample/2 sec, CSV file output Remote Chilled Mirror Hygrometer, Remote Mount, Tested up to 90°C at 95% RH, with Air Temperature Probe
Call For Price
Roscid Technologies DewTran® Temperature Probe for Chilled Mirror Hygrometer 7893056 Mfr # DewTran -T
COMPARE
SD 64GB, 1 sample/2 sec, CSV file output Remote Chilled Mirror Hygrometer with Air Temperature Probe
Call For Price
Roscid Technologies DewTran® Chilled Mirror Hygrometer, Remote Mount 7893057 Mfr # DewTan-R
COMPARE
SD 64GB, 1 sample/2 sec, CSV file output Remote Chilled Mirror Hygrometer, Remote Mount
Call For Price
Showing 1- 9 of 9