Roscid Technologies CM10 Chilled Mirror Hygrometers
Single-stage chilled mirror hygrometer for duct mount, wall mount, and OEM applications
  • Precise and accurate measurements Dual 4-20 mA configurable outputs Robust durability Swift performance Versatile duct, pipe, and wall mount configurations Automatic balance function for the mirror/optical loop circuitry
The CM10 single-stage chilled mirror hygrometers offer stable, accurate, and repeatable measurements that are traceable to NIST. Its simple construction allows for a long service life with excellent performance. Ideal for use in HVAC, clean room, environmental chamber, pharmaceutical, industrial process, glovebox, semiconductor, data storage, hospital, museum, and battery system applications.
The CM10 requires 24VDC at 1 A for power. It has two 4-20 mA outputs factory scaled for -40°C to 60°C. The unit also has an RS-232 communication connection that can be accessed using either the Hyper terminal or Tera Term program which is available on most computers. Service cycles for balancing and cleaning the mirror can be set on the unit. Mirror cleaning is quick and easy, needing only a wet cotton swab to restore the transmitter to normal operation.
Call For Price
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Max Dew Point Range (° C) Min Dew Point Range (° C) Mounting Type
Availability Pricing
Roscid Technologies CM10 Series Duct Mount Single-Stage Chilled Mirror Hygrometer 7893043 Mfr # CM10-D
COMPARE
60 -20 Duct
Call For Price
Roscid Technologies CM10 Series Wall Mount Single-Stage Chilled Mirror Hygrometer 7893044 Mfr # CM10-W
COMPARE
60 -20 Wall
Call For Price
Roscid Technologies CM10 Series Wall Mount Single-Stage Chilled Mirror Hygrometer with Temperature 7893045 Mfr # CM10-W-T
COMPARE
60 -20 Wall
Call For Price
Roscid Technologies CM10 Series OEM Configuration (No Enclosure) Single-Stage Chilled Mirror Hygrometer 7893046 Mfr # CM10-OEM
COMPARE
60 -20 OEM
Call For Price
Roscid Technologies CM10 Series OEM Configuration (No Enclosure) Single-Stage Chilled Mirror Hygrometer with Temperature 7893047 Mfr # CM10-OEM-T
COMPARE
60 -20 OEM
Call For Price
Roscid Technologies CM10 Series Pipe Mount Single-Stage Chilled Mirror Hygrometer 7893048 Mfr # CM10-P
COMPARE
60 -20 Pipe