Revolutionary Science Incufridge Series 300 Refrigerated Incubators
Affordable option for your heating and cooling needs
  • Compact design
  • Easy calibration
  • Refrigerant-free and Peltier based
  • Thick, closed cell insulation
  • Rapid ramp rate to set point
  • ISO 9001 Registered
The 300 series refrigerated incubators are a reliable and affordable option for your heating and cooling needs. Thermostatically controlled, these Peltier-based, refrigerant-free incubators are flexible when refrigerating or incubating temperature sensitive material. Each model is insulated with polystyrene foam and features a stainless-steel chamber and rugged powder-coated steel external construction.
The professional models (31506-01 & -03) include a keyed lock, clear inner door for sample monitoring, rear passthrough port, and color touch screen display. Features include advanced temperature programming, built-in data logging, and WiFi connectivity for remote management and alert notifications.
Ideal for standard and advanced lab requirements. Applications included DNA storage, elegans reproduction, BOD water analysis, yeast growth, bacterial proliferation, and spore testing.
£1,710.57 - £3,394.57GBP / Each
£2,052.68 - £4,073.48GBP (incl VAT)
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (Liters) Max Temperature (° C) Number Of Shelves
Availability Pricing
Revolutionary Science Incufridge 328S Standard Refrigerated Incubator, 28L; 120V WZ-31506-00 Mfr # RS-IF-328S
COMPARE
28 60 1
£1,710.57GBP / Each
£2,052.68GBP (incl VAT)
Revolutionary Science Incufridge 328P Professional Refrigerated Incubator, 28L; 120V WZ-31506-01 Mfr # RS-IF-328P
COMPARE
28 75 2
£2,356.82GBP / Each
£2,828.18GBP (incl VAT)
Revolutionary Science Incufridge 365P Professional Refrigerated Incubator, 65L; 120V WZ-31506-03 Mfr # RS-IF-365P
COMPARE
65 75 2
£3,394.57GBP / Each
£4,073.48GBP (incl VAT)