Replacement Spindles for Brookfield Rotational Viscometers
Order replacement spindles according to your viscometer model and desired centipoise range.
£226.49 - £631.56GBP / Each
£271.79 - £757.87GBP (incl VAT)
11 variations of this product are available.
See All Product Options Below
11 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Viscosity Range (cP)
Availability Pricing
Brookfield AMETEK LV-1 Replacement spindle for models 98945-00, -05, 15 to 20,000 cp 9894570 Mfr # LV-1
COMPARE
20000
£509.60GBP / Each
£611.52GBP (incl VAT)
Brookfield AMETEK LV-2 Replacement spindle for models 98945-00, -05, 50 to 100,000 cp 9894571 Mfr # LV-2
COMPARE
100000
£276.15GBP / Each
£331.38GBP (incl VAT)
Brookfield AMETEK LV-3 Replacement spindle for models 98945-00, -05, 200 to 400,000 cp 9894572 Mfr # LV-3
COMPARE
400000
Call For Price
Brookfield AMETEK LV-4 Replacement spindle for models 98945-00, -05, 1000 to 2,000,000 cp 9894573 Mfr # LV-4
COMPARE
2000000
£235.66GBP / Each
£282.79GBP (incl VAT)
Brookfield AMETEK RV-1 Replacement Viscometer Spindle 9894574 Mfr # RV-1
COMPARE
160000
£631.56GBP / Each
£757.87GBP (incl VAT)
Brookfield AMETEK RV-2 Replacement Viscometer Spindle 9894575 Mfr # RV-2
COMPARE
640000
£272.66GBP / Each
£327.19GBP (incl VAT)
Brookfield AMETEK RVHV-3 Replacement Viscometer Spindle 9894576 Mfr # RVHV-3
COMPARE
1600000
£316.22GBP / Each
£379.46GBP (incl VAT)
Brookfield AMETEK RVHV-4 Replacement Viscometer Spindle 9894577 Mfr # RVHV-4
COMPARE
3200000
£278.14GBP / Each
£333.77GBP (incl VAT)
Brookfield AMETEK RVHV-5 Replacement Viscometer Spindle 9894578 Mfr # RVHV-5
COMPARE
6400000
£226.49GBP / Each
£271.79GBP (incl VAT)
Brookfield AMETEK RVHV-6 Replacement Viscometer Spindle 9894579 Mfr # RVHV-6
COMPARE
16000000
£271.79GBP / Each
£326.15GBP (incl VAT)
Brookfield AMETEK RVHV-7 Replacement Viscometer Spindle 9894580 Mfr # RVHV-7
COMPARE
64000000
£316.22GBP / Each
£379.46GBP (incl VAT)
Showing 1- 11 of 11