Reagents® Cupric Sulfate Pentahydrate, ACS Reagent

Manufactured using the highest quality raw materials in ISO 9001 facility
  • Minimum Assay 98%
  • Blue solid
  • Density: 2.29
  • Boiling point: >150 °C (302 °F)
  • Solubility information: 220 g/L at 25 °C (77 °F)
Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.
Call For Price

2 variations of this product are available.

See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Class Packaging Weight (g)
Availability Pricing
Reagents Cupric Sulfate Pentahydrate, ACS Reagent, Natural Poly Container, 125 g 6228167 Mfr # C1190500-125A6
COMPARE
Salts Natural poly container 125
Call For Price
Reagents Cupric Sulfate Pentahydrate, ACS Reagent, Poly Pail, 12 kg 6228168 Mfr # C1190500-12KG
COMPARE
Salts Poly pail 12000
Call For Price
Showing 1- 2 of 2

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At+44 (0)1480 272279
Live chat