Reagents® Bromophenol Blue, ACS Reagent

Manufactured using the highest quality raw materials in ISO 9001 facility
  • Pale orange solid
  • Density: 0.95
  • Solubility nformation: 4 g/L at 20 °C (68 °F)
  • pH range: 3.0 (yellow) to 4.6 (blue)
Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.
Call For Price

2 variations of this product are available.

See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Class Packaging Weight (g)
Availability Pricing
Reagents Bromophenol Blue, ACS Reagent, Wide-Mouth Amber Glass Bottle, 10 g 6228660 Mfr # C6106500-10C1
COMPARE
Indicator Wide-mouth amber glass bottle 10
Call For Price
Reagents Bromophenol Blue, ACS Reagent, Wide-Mouth Amber Glass Bottle, 5 g 6228661 Mfr # C6106500-5C2
COMPARE
Indicator Wide-mouth amber glass bottle 5
Call For Price
Showing 1- 2 of 2

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At+44 (0)1480 272279
Live chat