Reagents® Barium Chloride, ACS Reagent

Manufactured using the highest quality raw materials in ISO 9001 facility
  • Colorless/white solid
  • Density: 3.86
  • Solubility information: 370 g/L at 20 °C (68 °F)
  • pH range: 5.2 to 8.2
Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.
Call For Price

4 variations of this product are available.

See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Class Packaging Weight (g)
Availability Pricing
Reagents Barium Chloride Dihydrate, ACS Reagent, Wide-Mouth Container, 100 g 6228154 Mfr # C1147000-100A6
COMPARE
Salts Wide-mouth container 100
Call For Price
Reagents Barium Chloride Dihydrate, ACS Reagent, Poly Pail, 10 kg 6228155 Mfr # C1147000-10KG
COMPARE
Salts Poly pail 10000
Call For Price
Reagents Barium Chloride Dihydrate, ACS Reagent, Natural Poly Container, 3 kg 6228156 Mfr # C1147000-3A5
COMPARE
Salts Poly container 3000
Call For Price
Reagents Barium Chloride Dihydrate, ACS Reagent, Fiber Drum, 50 kg 6228157 Mfr # C1147000-50KG
COMPARE
Salts Fiber drum 50000
Call For Price
Showing 1- 4 of 4

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At+44 (0)1480 272279
Live chat