QLA Pharmatest-Compatible Dissolution Vessels
QLA vessels provide more precise and consistent dissolution results
  • 1000 mL capacity
Vessels are manufactured using a unique molding process to provide a precise and consistent inside diameter and spherical radius to meet the physical dimensions of the USP.
Standard vessels are manufactured to meet USP physical dimensions and have a perfectly rounded hemispherical bottom. Footed plastic vessels are an economical alternative to glass. They feature feet which allow them to stand upright independently.
£131.57 - £232.57GBP / Each
£157.88 - £279.08GBP (incl VAT)
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Bottle Color Material Special Features
Availability Pricing
QLA Standard Vessel for Pharmatest, 1000 mL, Amber Glass; Serialized WZ-41306-26 Mfr # GLA900-APT
COMPARE
Amber Glass Standard
£232.57GBP / Each
£279.08GBP (incl VAT)
QLA Standard Vessel for Pharmatest, 1000 mL, Clear Glass; Serialized WZ-41306-38 Mfr # GLA900-PT
COMPARE
Clear Glass Footed
£168.92GBP / Each
£202.70GBP (incl VAT)
QLA Footed Vessel for Pharmatest, 1000 mL; Amber Plastic WZ-41306-59 Mfr # PLA900-AP
COMPARE
Amber Plastic Standard
£145.13GBP / Each
£174.16GBP (incl VAT)
QLA Footed Vessel for Pharmatest, 1000 mL; Clear Plastic WZ-41306-64 Mfr # PLA900-PT
COMPARE
Clear Plastic Footed
£131.57GBP / Each
£157.88GBP (incl VAT)