PharmaPure® Low Spallation Peristaltic Pump Tubing
Low spallation, biologically compatible tubing—developed specially for pharmaceutical, biotechnology, and laboratory applications
  • Durable tubing provides long pump life, outlasting silicone tubing
  • Excellent gas barrier with very low gas permeability
  • Withstands repeated autoclaving
  • USP Class VI and European Pharmacopoeia 3.2.9 compliant
  • Ideal for general-purpose, laboratory, and single-use applications
  • Free technical application support available to assist with tubing selection
Made from a proprietary multilayer thermoplastic elastomer, PharmaPure peristaltic pump tubing offers excellent pump life, ultra-low particulate spallation, and very low gas permeability. Developed especially for the pharmaceutical and biotechnology industries, it is ideal for use in media processing, vaccine manufacturing, aseptic filling, and diagnostic testing applications.
Exceptional flex life characteristics and excellent wear properties of the PharmaPure tubing contribute to a long service life in peristaltic pumps and help minimize downtime due to tubing failures. The low permeability and low absorption characteristics ensure sensitive fluids are protected from gas ingress and concentration changes due to fluid absorption.
£93.78 - £483.71GBP / Pkg of 1
£112.54 - £580.45GBP (incl VAT)
13 variations of this product are available.
See All Product Options Below
13 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Tubing ID (in) Tubing OD (in) Wall Thickness (in)
Availability Pricing
Pharmapure Low Spallation Peristaltic Pump Tubing, 1/32" ID x 5/32" OD; 25 Ft 5012293 Mfr # AL242606
COMPARE
1/32 5/32 1/16
£93.78GBP / Pkg of 1
£112.54GBP (incl VAT)
Pharmapure Low Spallation Peristaltic Pump Tubing, 1/16" ID x 1/8" OD; 25 Ft 5012294 Mfr # AL242002
COMPARE
1/16 1/8 1/32
£103.94GBP / Pkg of 1
£124.73GBP (incl VAT)
Pharmapure Low Spallation Peristaltic Pump Tubing, 1/16" ID x 3/16" OD; 25 Ft 5012295 Mfr # AL242003
COMPARE
1/16 3/16 1/16
£115.66GBP / Pkg of 1
£138.79GBP (incl VAT)
Pharmapure Low Spallation Peristaltic Pump Tubing, 1/8" ID x 1/4" OD; 25 Ft 5012296 Mfr # AL242007
COMPARE
1/8 1/4 1/16
£120.35GBP / Pkg of 1
£144.42GBP (incl VAT)
Pharmapure Low Spallation Peristaltic Pump Tubing, 3/16" ID x 5/16" OD; 25 Ft 5012297 Mfr # AL242012
COMPARE
3/16 5/16 1/16
£122.69GBP / Pkg of 1
£147.23GBP (incl VAT)
Pharmapure Low Spallation Peristaltic Pump Tubing, 1/4" ID x 3/8" OD; 25 Ft 5012298 Mfr # AL242017
COMPARE
1/4 3/8 1/16
£144.96GBP / Pkg of 1
£173.95GBP (incl VAT)
Pharmapure Low Spallation Peristaltic Pump Tubing, 1/4" ID x 1/2" OD; 25 Ft 5012299 Mfr # AL242019
COMPARE
1/4 1/2 1/8
£254.75GBP / Pkg of 1
£305.70GBP (incl VAT)
Pharmapure Low Spallation Peristaltic Pump Tubing, 5/16" ID x 7/16" OD; 25 Ft 5012300 Mfr # AL242022
COMPARE
5/16 7/16 1/16
£151.99GBP / Pkg of 1
£182.39GBP (incl VAT)
Pharmapure Low Spallation Peristaltic Pump Tubing, 3/8" ID x 1/2" OD; 25 Ft 5012301 Mfr # AL242027
COMPARE
3/8 1/2 1/16
£185.99GBP / Pkg of 1
£223.19GBP (incl VAT)
Pharmapure Low Spallation Peristaltic Pump Tubing, 3/8" ID x 5/8" OD; 25 Ft 5012302 Mfr # AL242029
COMPARE
3/8 5/8 1/8
£240.69GBP / Pkg of 1
£288.83GBP (incl VAT)
Pharmapure Low Spallation Peristaltic Pump Tubing, 1/2" ID x 3/4" OD; 25 Ft 5012303 Mfr # AL242038
COMPARE
1/2 3/4 1/8
£293.83GBP / Pkg of 1
£352.60GBP (incl VAT)
Pharmapure Low Spallation Peristaltic Pump Tubing, 5/8" ID x 7/8" OD; 25 Ft 5012304 Mfr # AL242046
COMPARE
5/8 7/8 1/8
£346.96GBP / Pkg of 1
£416.35GBP (incl VAT)
Pharmapure Low Spallation Peristaltic Pump Tubing, 3/4" ID x 1" OD; 25 Ft 5012305 Mfr # AL242053
COMPARE
3/4 1 1/8
£483.71GBP / Pkg of 1
£580.45GBP (incl VAT)
Showing 1- 13 of 13