PharmaLok™ Tubing Clamps
Easily and securely close off flow in flexible tubing
  • Lightweight and reusable
  • Glass-filled nylon construction—rust and corrosion resistant
  • Easy, one-handed closure—no tools needed
  • Tamper evident and zip tie compatible
  • Sterilize by autoclave, gamma irradiation, or x-ray
  • FDA 21 CFR 177.1500, USP Class VI, and ISO 10993-5/10993-11 compliant
  • Animal-derived ingredient, BPA, and BSE/TSE free
  • Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system
  • Free technical application support available to determine which connector type is needed for your application
Effortlessly close off flow in tubing with PharmaLok tubing clamps. The open-clamp style allows it to be easily installed on existing tubing assemblies. These simple-to-use tubing clamps require no tools and can be operated using one hand. Clamps accept a wide range of flexible tubing materials and sizes.
£285.78 - £385.78GBP / Pkg of 10
£342.94 - £462.94GBP (incl VAT)
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Clamp Material Max Grip Range (InchShort) Description
Availability Pricing
PharmaLok Small and Thin-Walled Tubing Clamp, 1-1/8" OD Max; 10/Pk 5013436 Mfr # CPT90-U71
COMPARE
Glass-filled nylon 1.125 PharmaLok™ Small and Thin-Walled Tubing Clamp, 1-1/8" OD Max; 10/Pk
£285.78GBP / Pkg of 10
£342.94GBP (incl VAT)
PharmaLok Standard Tubing Clamp, 1-1/4" OD Max; 10/Pk 5013437 Mfr # CPT100-U71
COMPARE
Glass-filled nylon 1.25 PharmaLok™ Standard Tubing Clamp, 1-1/4" OD Max; 10/Pk
£295.56GBP / Pkg of 10
£354.67GBP (incl VAT)
PharmaLok Large Tubing Clamp, 2" OD Max; 10/Pk 5013438 Mfr # CPT110-U71
COMPARE
Glass-filled nylon 2 PharmaLok™ Large Tubing Clamp, 2" OD Max; 10/Pk
£385.78GBP / Pkg of 10
£462.94GBP (incl VAT)