PendoTECH® Transmitters for Single-Use Temperature Sensors
Convert temperature measurements from single-use temperature sensors to 4 to 20 mA output signals
  • Linear 4 to 20 mA output signal reflects 0 to 70°C (32°F to 158°F), the measuring range of the sensor
  • Rugged ABS plastic enclosure offers durability
Connect a single-use temperature sensor to the transmitter to convert the temperature measurement to a linear 4 to 20 mA output signal. Simply plug the sensor to a reusable sensor cable 48524-82 for a quick and easy connection to the transmitter. Use interface cables 48524-85 or -86 to transmit a 4 to 20 mA output signal from the transmitter to PressureMAT® Plus monitor for real-time temperature monitoring.
£1,103.41GBP / Each
£1,324.09GBP (incl VAT)
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Number of Channels
Availability Pricing
PendoTech Benchtop Transmitter for Single-Use Temperature Sensor, 1 Input 4852483 Mfr # PDKT-TT1
COMPARE
1
£1,103.41GBP / Each
£1,324.09GBP (incl VAT)