PendoTECH® Single-Use Temperature Sensors™
Accurate, cost-effective, and single use for elimination of cross-contamination
  • No calibration required—each sensor is pretested for accuracy
  • High sensitivity with better than ±0.2°C accuracy in the 0 to 70°C range
  • Biocompatible materials meeting USP Class VI standards
  • Gamma and x-ray irradiation compatible for sterilization
PendoTECH® Single-Use Temperature Sensors™ deliver precise and reliable in-line temperature measurements for bioprocess applications such as filtration, chromatography, filling operations, and general process monitoring. Connect sensors to a transmitter via a reusable cable; transmitter provides a 4 to 20 mA output signal for an easy connection to PressureMAT® Plus for real-time monitoring (interface cable required).
Sensors feature hose barb fittings to prevent pressure drops and dead legs, and require no calibration, ensuring easy setup and dependable performance. Designed to minimize contamination risks while maintaining high accuracy (±0.2°C or better), they are available in multiple sizes to integrate smoothly into existing workflows. Manufactured in an ISO 9001-certified facility, these sensors provide a cost-effective alternative to traditional temperature monitoring devices while supporting single-use bioprocessing needs.
£67.71 - £89.45GBP / Each
£81.25 - £107.34GBP (incl VAT)
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Process Connection Sterile Wetted Materials
Availability Pricing
PendoTech Single-Use Temperature Sensor, Non-Sterile, Polysulfone, 1/8" Hose Barb 5990023 Mfr # TEMPS-N-012
COMPARE
1/8" Hose Barb No Polysulfone
£89.45GBP / Each
£107.34GBP (incl VAT)
PendoTech Single-Use Temperature Sensor, Non-Sterile, Polysulfone, 1/4" Hose Barb 5990024 Mfr # TEMPS-N-025
COMPARE
1/4" Hose Barb No Polysulfone
£71.56GBP / Each
£85.87GBP (incl VAT)
PendoTech Single-Use Temperature Sensor, Non-Sterile, Polysulfone, 3/8" Hose Barb 5990025 Mfr # TEMPS-N-038
COMPARE
3/8" Hose Barb No Polysulfone
£73.93GBP / Each
£88.72GBP (incl VAT)
PendoTech Single-Use Temperature Sensor, Non-Sterile, Polysulfone, 1/2" Hose Barb 5990026 Mfr # TEMPS-N-050
COMPARE
1/2" Hose Barb No Polysulfone
£67.71GBP / Each
£81.25GBP (incl VAT)