Orochem Reliasil™ C8 HPLC Columns
Rugged design with enhanced chemical stability and efficiency
  • Excellent mechanical and chemical stability
  • Ideal for reverse phase separations of peptides, pharmaceuticals, and derivatized amino acids
  • Suitable for C8 phase
The Orochem Reliasil HPLC columns are designed for high efficiency (high plate count) and general utility. The proprietary silica gel manufacturing process yields a Gaussian particle size distribution, narrow pore-size distribution, and strong resistance to high pH. All columns feature a 190 m²/g surface area and 0.55 pore volume. Choose from standard columns with a pore size of 90 Å or columns with a 300 Å pore size.
£569.96 - £5,392.60GBP / Each
£683.95 - £6,471.12GBP (incl VAT)
11 variations of this product are available.
See All Product Options Below
11 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Column Inside Diameter (ID) (mm) Column Length (mm) Column Particle Size (µm)
Availability Pricing
Orochem Reliasil™ HPLC Column, C8, 3 µm, 100 mm L x 3.0 mm ID, 90Å 0648659 Mfr # R3DG-102
COMPARE
3 100 3
£614.76GBP / Each
£737.71GBP (incl VAT)
Orochem Reliasil™ HPLC Column, C8, 5 µm, 100 mm L x 3.0 mm ID, 90Å 0648660 Mfr # R5DG-122
COMPARE
3 100 5
£569.96GBP / Each
£683.95GBP (incl VAT)
Orochem Reliasil™ HPLC Column, C8, 5 µm,  150 mm L x 4.6 mm ID, 300Å 0648661 Mfr # R5FI-127
COMPARE
4.6 150 5
£696.89GBP / Each
£836.27GBP (incl VAT)
Orochem Reliasil™ HPLC Column, C8, 3 µm,  150 mm L x 4.6 mm ID, 90Å 0648662 Mfr # R3FI-102
COMPARE
4.6 150 3
£589.04GBP / Each
£706.85GBP (incl VAT)
Orochem Reliasil™ HPLC Column, C8, 5 µm,  150 mm L x 4.6 mm ID, 90Å 0648663 Mfr # R5FI-122
COMPARE
4.6 150 5
£589.04GBP / Each
£706.85GBP (incl VAT)
Orochem Reliasil™ HPLC Column, C8, 5 µm,  250 mm L x 4.6 mm ID, 300Å 0648664 Mfr # R5HI-127
COMPARE
4.6 250 5
£783.18GBP / Each
£939.82GBP (incl VAT)
Orochem Reliasil™ HPLC Column, C8, 5 µm,  2500 mm L x 4.6 mm ID, 90Å 0648665 Mfr # R5HI-122
COMPARE
4.6 250 5
£673.66GBP / Each
£808.39GBP (incl VAT)
Orochem Reliasil™ HPLC Column, C8, 10 µm, 250 mm L x 10 mm ID, 90Å 0648666 Mfr # R9HL-145
COMPARE
10 250 10
£2,970.08GBP / Each
£3,564.10GBP (incl VAT)
Orochem Reliasil™ HPLC Column, C8, 5 µm,  250 mm L x 10 mm ID, 90Å 0648667 Mfr # R5HL-145
COMPARE
10 250 5
£2,837.34GBP / Each
£3,404.81GBP (incl VAT)
Orochem Reliasil™ HPLC Column, C8, 10 µm, 250 mm L x 21 mm ID, 90Å 0648668 Mfr # R9HM-145
COMPARE
21 250 10
£5,392.60GBP / Each
£6,471.12GBP (incl VAT)
Orochem Reliasil™ HPLC Column, C8, 5 µm, 250 mm L x 21 mm ID, 90Å 0648669 Mfr # R5HM-145
COMPARE
21 250 5
£3,173.34GBP / Each
£3,808.01GBP (incl VAT)
Showing 1- 11 of 11