Ohaus Moisture Balances
Innovative carbon fiber heating enables faster analysis
  • Upgraded load cell offers accurate and consistent results
  • Intuitive operation system provides convenient testing
  • Enhanced stability reduces vibrations and temperature fluctuations
  • Robust design ensures enhanced durability and stability
  • Overload protection reduces risk of damage
  • Easy cleaning and maintenance
These next generation of MB series moisture balances come with a new carbon fiber heating technology and upgraded loadcell for shorten analysis time, better power saving, and accurate and consistent results. Ideal for various applications, including food, pharmaceutical, and chemical industries.
The robust, sleek design offers improved durability and stability in harsh environments. The moisture balances are easy to operate with intuitively placed navigation control and accessible sample chamber with soft-close lid. Smooth surfaces and easily accessible components simplify cleaning and maintenance routines, ensuring the balance remains in optimal condition.
Customizable parameters allow you to adjust various settings such as temperature profiles, drying times, and endpoint criteria to suit specific sample types and analysis requirements. The balances allow you to save and quickly recall multiple analysis methods, allowing for efficient switching between different tests without needing to reconfigure the device each time. A user-friendly interface makes it easier for both novice and experienced users to optimize their analyses.
£2,947.68 - £4,376.30GBP / Each
£3,537.22 - £5,251.56GBP (incl VAT)
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (g) Heater Type Readability (g)
Availability Pricing
Ohaus MB32 Moisture Balance, 90g x 1mg, 4" LCD, 2 Drying Programs 1160103 Mfr # MB32
COMPARE
90 Carbon fiber 0.001
£2,947.68GBP / Each
£3,537.22GBP (incl VAT)
Ohaus MB62 Moisture Balance, 90g x 1 mg, 5" LCD, 4 Drying Programs 1160104 Mfr # MB62
COMPARE
90 Carbon fiber 0.001
£3,750.76GBP / Each
£4,500.91GBP (incl VAT)
Ohaus MB92 Moisture Balance, 90g x 1mg, Color Touchscreen, 4 Drying Programs 1160105 Mfr # MB92
COMPARE
90 Carbon fiber 0.001
£4,376.30GBP / Each
£5,251.56GBP (incl VAT)