Ohaus® Incubating Rocking and Waving Shakers
Specially designed to provide a smooth rocking or waving motion while simultaneously incubating samples
  • Control and view speed/tilt, temperature, and time independently
  • Precise adjustments of tilt angle, speed, and temperature
  • View actual/setpoint temperatures
  • Count up/count down timer
Use the incubating rocking shaker and incubating waving shaker in hybridization, cell culture, blotting, staining, and destaining applications. Both models feature independent controls for speed, temperature, and time with an intuitive touchpad, making it easy to set and view the parameters. Make tilt angle, speed, and temperature adjustments while in operation. Adjust tilt angle in 1° increments, vary speed by 1 rpm, and set temperature in 1 °C increments. The speed ramping feature increases speed slowly to avoid splashing.
Set the temperature from ambient +5 °C to 65 °C (149 °F); a polycarbonate dome cover helps maintain uniform chamber temperature. Built with operator safety in mind, a “Caution Hot” indicator illuminates if the chamber air temperature is above 40 °C (104 °F). Set the timer to display accumulated time (continuous mode) or remaining time (timed mode) from 1 second to 9999 minutes.
£3,228.09 - £3,298.61GBP / Each
£3,873.71 - £3,958.33GBP (incl VAT)
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Speed (rpm) Shaking Angle Description
Availability Pricing
Ohaus Incubating Rocking Shaker, 1 to 50 rpm, 0 to 15° Tilt 5302780 Mfr # ISRK04HDG
COMPARE
50 0 to 15° Incubating Rocking Shaker, 1 to 50 rpm, 0 to 15° Tilt
£3,298.61GBP / Each
£3,958.33GBP (incl VAT)
Ohaus Incubating Waving Shaker, 1 to 30 rpm, 0 to 20° Tilt 5302781 Mfr # ISWV02HDG
COMPARE
30 0 to 20° Incubating Waving Shaker, 1 to 30 rpm, 0 to 20° Tilt
£3,228.09GBP / Each
£3,873.71GBP (incl VAT)