Ohaus® Frontier™ Multi Centrifuge
Compact, low-speed centrifuge accepts up to 50 mL capacity tubes
  • Holds conical and round-bottom tubes without adapter
  • Tool-free rotor change
  • Large LCD with simple interface
  • Easily switch between rpm and RCF
  • 10 acceleration/deceleration settings
  • Preset run time from 10 seconds to 99 hours 59 minutes or continuous
  • Mechanical locking system ensures lid stays shut while in operation
  • Imbalance sensor stops rotor when excess force is detected
Use this small benchtop centrifuge for low-speed centrifugation applications, such as cell culture, blood separation, and sample prep for flow cytometry. Quickly and easily install and remove rotors without the use of any tools. The versatile centrifuge accommodates all standard tube sizes from 1.5 to 50 mL; hold up to six 50 mL tubes or twelve 15 mL tubes at once.
View actual and set readings on the large backlit LCD. Easy-to-turn rotary knob offers quick parameter adjustments. Select from 10 acceleration and 10 deceleration rates to suit your application. Safety features include a mechanical lid-locking system that prevents the lid from opening during operation and an imbalance sensor that stops the rotor when imbalanced load is detected.
£1,286.25 - £1,680.00GBP / Each
£1,543.50 - £2,016.00GBP (incl VAT)
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Capacity (mL) Max Speed (rpm) Description
Availability Pricing
Ohaus Frontier FC5706P Multi-Function Centrifuge, 6000 rpm, 100-230V 8524100 Mfr # 31060778
COMPARE
50 6000 Multi-Function Centrifuge, 6000 rpm, 100-230V
£1,680.00GBP / Each
£2,016.00GBP (incl VAT)
Ohaus Frontier FC5707 Low-Speed Multi Centrifuge with Touch-Wheel Control, 6800 rpm, Bundle with 8 x 15 ml Rotor, 100-230V 8524103 Mfr # 30393189
COMPARE
15 6800 Low-Speed Multi Centrifuge with Touch-Wheel Control, 6800 rpm, Bundle with 8 x 15 ml Rotor, 100-230V
£1,438.50GBP / Each
£1,726.20GBP (incl VAT)
Ohaus FrontierFC5707 Low-Speed Multi Centrifuge with Touch-Wheel Control, 6800 rpm, Bundle with 4 x 50ml RB/FA or 4 x 15ml RB/FA Rotor, 100-230V 8524104 Mfr # 83041073
COMPARE
50 6800 Low-Speed Multi Centrifuge with Touch-Wheel Control, 6800 rpm, Bundle with 4 x 50ml RB/FA or 4 x 15ml RB/FA Rotor, 100-230V
£1,286.25GBP / Each
£1,543.50GBP (incl VAT)