Ohaus® Frontier™ High-Speed Micro Centrifuges
HIgh-speed benchtop micro centrifuges accept a broad range of accessories adding versatility
  • Compact design saves valuable bench space in the lab
  • View actual and set readings on the large backlit LCD
  • Easily maneuver controls even while wearing gloves
  • 10 acceleration/deceleration settings
  • Preset run time from 10 seconds to 99 hours 59 minutes or continuous
  • 99 memory sets to quickly store and recall commonly used parameters
  • Motorized lid-locking system prevents the lid from opening during operation
  • Imbalance sensor stops the rotor when imbalance is detected
The versatile and easy-to-use Frontier benchtop micro centrifuges deliver high speeds, making them ideal for DNA sample preparation, protein separation, and cell pelleting. Quickly make parameter adjustments using a touch wheel or rotary knob. Select from 10 acceleration and 10 deceleration rates to suit your application. Store up to 99 runs with relevant parameters, including the used rotors, to easily repeat the procedures and save time. Safety features include a motorized lid-locking system that prevents the lid from opening during operation and an imbalance sensor that stops the rotor when imbalanced load is detected.
£1,779.75 - £6,740.75GBP / Each
£2,135.70 - £8,088.90GBP (incl VAT)
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Speed (rpm) Refrigerated Description
Availability Pricing
Ohaus Frontier FC5513  High-Speed Micro Centrifuge with Touch-Wheel Control, 13,500 rpm, 120V 8524001 Mfr # 30393188
COMPARE
13500 No High-Speed Micro Centrifuge with Touch-Wheel Control, 13,500 rpm, 120V
£2,251.62GBP / Each
£2,701.94GBP (incl VAT)
Ohaus Frontier FC5515 High-Speed Micro Centrifuge with Rotary Knob Control, 15,200 rpm, 120V 8524002 Mfr # 30130867
COMPARE
15200 No High-Speed Micro Centrifuge with Rotary Knob Control, 15,200 rpm, 120V
£3,087.06GBP / Each
£3,704.47GBP (incl VAT)
Ohaus Frontier FC5515R High-Speed Refrigerated Micro Centrifuge with Rotary Knob Control, 15,200 rpm, 120V 8524003 Mfr # 83042058
COMPARE
15200 Yes High-Speed Refrigerated Micro Centrifuge with Rotary Knob Control, 15,200 rpm, 120V
£6,740.75GBP / Each
£8,088.90GBP (incl VAT)
Ohaus Frontier FC5513 High-Speed Micro Centrifuge with Touch-Wheel Control, 13,500 rpm, 230V 8524097 Mfr # 30393187
COMPARE
13500 No High-Speed Micro Centrifuge with Touch-Wheel Control, 13,500 rpm, 230V
£1,779.75GBP / Each
£2,135.70GBP (incl VAT)
Ohaus Frontier FC5515 High-Speed Micro Centrifuge with Rotary Knob Control, 15,200 rpm, 230V 8524098 Mfr # 30130866
COMPARE
15200 No High-Speed Micro Centrifuge with Rotary Knob Control, 15,200, 230V
£1,963.50GBP / Each
£2,356.20GBP (incl VAT)
Ohaus Frontier FC5515R High-Speed Refrigerated Micro Centrifuge with Rotary Knob Control, 15,200 rpm, 230V 8524099 Mfr # 83042057
COMPARE
15200 Yes High-Speed Refrigerated Micro Centrifuge with Rotary Knob Control, 15,200 rpm, 230V
£5,034.75GBP / Each
£6,041.70GBP (incl VAT)
Ohaus Frontier FC5513 High Speed Micro Centrifuge with Touch-Wheel Control, 14,000 rpm, Bundle with  18 x 1.5/2.0 ml Rotor, 120V 8524105 Mfr # 83041044
COMPARE
14000 No High Speed Micro Centrifuge with Touch-Wheel Control, 14,000 rpm, Bundle with 18 x 1.5/2.0 ml Rotor, 120V
£1,835.98GBP / Each
£2,203.18GBP (incl VAT)
Ohaus Frontier FC5513 High Speed Micro Centrifuge with Touch-Wheel Control, 13,500 rpm, Bundle with 24 x 1.5/2.0 ml Rotor, 120V 8524106 Mfr # 30370692
COMPARE
13500 No High Speed Micro Centrifuge with Touch-Wheel Control, 13,500 rpm, Bundle with 24 x 1.5/2.0 ml Rotor, 120V
£2,440.78GBP / Each
£2,928.94GBP (incl VAT)
Showing 1- 8 of 8