NSI Lab Solutions Pharmaceutical Secondary Reference Materials, M to Z

Highly purified compounds—excellent for establishing qualitative and quantitative data
  • Direct traceability of purity and identity to pharmacopeia primary standards established by multiple methods of analysis
  • Great substitute for primary standards for routine quality control purposes
  • ISO 17034 accredited
Pharmaceutical secondary reference materials have been identity and purity certified against multiple analytical methods, including HPLC, FTIR, GC, GC/MS, melting point, and titration. See the Certificate of Analysis for details of the certification process used.

Download Certificate of Analysis at nsilabsolutions.com/coa-downloads/.
Call For Price

7 variations of this product are available.

See All Product Options Below
7 PRODUCT OPTIONS
Compareitem specifications
Element/Ion Description
Availability Pricing
COMPARE
2-Propanol 2-Propanol Pharmaceutical Reference Material, 1.2 mL; 3/PK
Call For Price
COMPARE
Sucrose Sucrose Pharmaceutical Reference Material; 1 g
Call For Price
COMPARE
Sucrose Sucrose Optical Rotation Pharmaceutical Reference Material; 50 g
Call For Price
COMPARE
Succinic Acid Succinic Acid Pharmaceutical Reference Material; 1 g
Call For Price
COMPARE
Sorbitol Sorbitol Pharmaceutical Reference Material; 1 g
Call For Price
COMPARE
Magnesium Carbonate Magnesium Carbonate Pharmaceutical Reference Material; 1 g
Call For Price
COMPARE
Methanol Methanol Pharmaceutical Reference Material; 20 mL
Call For Price
Showing 1- 7 of 7

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At+44 (0)1480 272279
Live chat