Lovibond Compact Color Meter
Attach text or an audio recording to your results for easy identification
  • Illuminate your sample using eight visible and one UV wavelength for more accurate color measurement
  • Each measurement is a combination of 27 images, giving information on color and texture effects
This handheld color meter measures surface color on graphic arts, painted surfaces, plastics, textiles, and more. Compact size and rechargeable batteries let you use this unit in the lab or carry on the plant floor for quality control testing.
Store up to 350 sample measurements; attach a text tag or audio message to any sample for information or identification purposes. Onboard software allows download of data in a formatted report or as a Microsoft® Excel® data table. Calibrate your color meter quickly and easily with the built-in reference slider.
£3,853.63GBP / Each
£4,624.36GBP (incl VAT)
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Detector Type Test Parameters Wavelength(s) (nm)
Availability Pricing
Lovibond Color Meter for Surfaces, 4/8-mm measuring area 5973451 Mfr # 401100
COMPARE
£3,853.63GBP / Each
£4,624.36GBP (incl VAT)