Lascar Electronics Compact USB Data Loggers, Temperature and Humidity
Direct-to-USB connection and easy-to-use functionality
  • User-programmable alarm thresholds
  • All data loggers come with software and are great for laboratory environmental monitoring
Simply insert any of these compact, 4.5" (11.4 cm) long loggers into your computer USB port, name it, choose your required sampling rate, select high and low alarms if required, and set the logger start time. Once configured, the logger can be removed from the computer to log independently. Data is downloaded back onto the PC where it can be analyzed and viewed as a graph using included software.
These data loggers measure temperature and humidity from 31 to 176°F (–35 to 80°C) and 0 to 100% RH, calculate dew point, and store over 16,000 readings per parameter.
£86.49 - £151.20GBP / Each
£103.79 - £181.44GBP (incl VAT)
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Temperature (° F) Temperature Accuracy (° F) Temperature Resolution (° F)
Availability Pricing
Lascar Electronics EL-USB-2 USB Temperature/Humidity Data Logger, 16,000+ readings, 1 year battery life 2303952 Mfr # EL-USB-2
COMPARE
176 ±0.9 1
£86.49GBP / Each
£103.79GBP (incl VAT)
Lascar Electronics EL-USB-2-LCD Data Logger, RH/Temp/Dew Pt, with LCD Display 2303962 Mfr # EL-USB-2-LCD
COMPARE
158 ±2.5F 1
£118.02GBP / Each
£141.62GBP (incl VAT)
Lascar Electronics EL-USB-2-LCD+ High Accuracy Data Logger, RH/Temp/Dew Pt, with LCD Display 2303965 Mfr # EL-USB-2-LCD+
COMPARE
176 ±0.6 1
£151.20GBP / Each
£181.44GBP (incl VAT)
Lascar Electronics EL-USB-2+ High Accuracy Data Logger, RH/Temp/Dew Pt 2303967 Mfr # EL-USB-2+
COMPARE
176 ±0.6 1
£109.93GBP / Each
£131.92GBP (incl VAT)