Labconco Replacement HEPA Filters for XPert® Systems and FilterMate™ Exhausters
Ensure user safety with HEPA filters
  • Removes 99.99% of particles 0.3 µm or larger
Replace HEPA filters regularly to ensure excellent filtration, ensuring user safety.
£782.03 - £844.10GBP / Each
£938.44 - £1,012.92GBP (incl VAT)
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Compatible With
Availability Pricing
Labconco 3707900 Replacement HEPA Filter for Exhauster, 2-ft XPert® Systems & Stations, and TXE Enclosures 0910156 Mfr # 3707900
COMPARE
FilterMate™ exhauster, 2-ft XPert® systems & stations, and TXE enclosures
£782.03GBP / Each
£938.44GBP (incl VAT)
Labconco 3707901 Replacement HEPA filter for 3-ft and 6-ft XPert® Systems and Stations 3350652 Mfr # 3707901
COMPARE
3-ft and 6-ft XPert® systems and stations (2 required for 6-ft units)
£844.10GBP / Each
£1,012.92GBP (incl VAT)