Idex® Peristaltic Tubing Adapters
Connect 1/16" OD rigid/semi-rigid tubing to soft-wall tubing
  • Retainer sleeve holds tubing tightly in place
Peristaltic tubing adapters feature specially designed nose to allow soft tubing to fit over the nose then be held in place by the retainer sleeve. To connect peristaltic tubing to rigid tubing with a different OD, order a flangeless fitting with 1/4-28 flat-bottom configuration.
£70.47 - £79.45GBP / Each
£84.56 - £95.34GBP (incl VAT)
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Barb Size / Tubing Id (Port 1) (mm) Compression Size / Tube Od (Port 1) (mm) Orifice Dimensions (mm)
Availability Pricing
Idex Peristaltic Tubing Adapter, PEEK/ETFE, 0.030" Bore, up to 0.18" OD x 0.048" to 0.11" ID; 1/EA 0201439 Mfr # P-757
COMPARE
3.8 6.35 0.75
£79.45GBP / Each
£95.34GBP (incl VAT)
Idex Peristaltic Tubing Adapter, Large Bore, PEEK/ETFE, 0.070" Bore, 0.25" OD x 0.10 to 0.15" ID; 2/PK 0202381 Mfr # P-767
COMPARE
2.8 4.55 1.78
£70.47GBP / Pkg of 2
£84.56GBP (incl VAT)