Idex® Inlet Solvent Filter Kits
Filter out particulate matter from the solvents to protect your pump and other components from damage
  • Includes filters, fittings, and tubing for quick set up
Use the inlet filters, fittings and tubing in the kit to protect your pump and other components from damage by filtering out particulate matter from the solvent. Inlet filters are made of PTFE and stainless steel, disposable, and easy to change without tools.
£120.72 - £198.64GBP / Each
£144.86 - £238.37GBP (incl VAT)
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Pore Size (µm)
Availability Pricing
Idex Inlet Solvent Filter Kit, 1/4-28 Coned Port, PTFE/SS, 10 µm, 1/8" OD Tubing 4271100 Mfr # A-200
COMPARE
10
£198.64GBP / Each
£238.37GBP (incl VAT)
Idex Inlet Solvent Filter Kit, 1/4-28 Coned Port, PTFE/SS, 2 µm, 1/8" OD Tubing 4271101 Mfr # A-201
COMPARE
2
£120.72GBP / Each
£144.86GBP (incl VAT)