HTL Optipette Single-Channel Pipettors
Easily adjust ejector height
  • Ergonomic handle design
  • Easy to calibrate
  • Rotating pipetting push button prevents accidental volume change
Remarkably robust and durable in a sleek design, Optipette adjustable-volume pipettors offer optimal accuracy and precision. With a UV-resistant body and autoclavable bottom half, the soft spring system reduces pipetting forces while color-coded rings simplify volume identification. Additional spacers enable ejector height adjustment; user-calibration is easy thanks to a provided calibration key.
Call For Price
10 variations of this product are available.
See All Product Options Below
10 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Volume (µL) Min Volume (µL) Number Of Channels
Availability Pricing
HTL 5600 Optipette OP250 Single Channel Pipettor, Green; 50 to 250 µL 2455183 Mfr # 5600
COMPARE
250 50 1
Call For Price
HTL 5601 Optipette OP2 Single Channel Pipettor, Red; 0.2 to 2 µL 2455184 Mfr # 5601
COMPARE
2 0.2 1
Call For Price
HTL 5602 Optipette OP10 Single Channel Pipettor, Red; 0.5 to 10 µL 2455185 Mfr # 5602
COMPARE
10 0.5 1
Call For Price
HTL 5603 Optipette OP20 Single Channel Pipettor, Yellow; 2 to 20 µL 2455186 Mfr # 5603
COMPARE
20 2 1
Call For Price
HTL 5604 Optipette OP100 Single Channel Pipettor, Yellow; 10 to 100 µL 2455187 Mfr # 5604
COMPARE
100 10 1
Call For Price
HTL 5605 Optipette OP200 Single Channel Pipettor, Yellow; 20 to 200 µL 2455188 Mfr # 5605
COMPARE
200 20 1
Call For Price
HTL 5606 Optipette OP1000 Single Channel Pipettor, Blue; 100 to 1000 µL 2455189 Mfr # 5606
COMPARE
1000 100 1
Call For Price
HTL 5607 Optipette OP50 Single Channel Pipettor, Yellow; 5 to 50 µL 2455190 Mfr # 5607
COMPARE
50 5 1
Call For Price
HTL 5608 Optipette OP5000 Single Channel Pipettor, White; 500 to 5000 µL 2455191 Mfr # 5608
COMPARE
5000 500 1
Call For Price
HTL 5609 Optipette OP10000 Single Channel Pipettor, White; 1000 to 10000 µL 2455192 Mfr # 5609
COMPARE
10000 1000 1
Call For Price
Showing 1- 10 of 10