HTL Optipette 8-Channel Pipettors
Easily adjust ejector height
  • Ergonomic handle design
  • Easy to calibrate
  • Rotating pipetting push button prevents accidental volume change
Remarkably robust and durable in a sleek design, Optipette adjustable-volume pipettors offer optimal accuracy and precision. With a UV-resistant body and autoclavable bottom half, the soft spring system reduces pipetting forces while color-coded rings simplify volume identification. Additional spacers enable ejector height adjustment; user-calibration is easy thanks to a provided calibration key.
Call For Price
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Volume (µL) Min Volume (µL) Number Of Channels
Availability Pricing
HTL 5141 Optipette OP8-10 Multichannel Pipettor, Red, 8-Channel; 0.5 to 10 µL 2455171 Mfr # 5141
COMPARE
10 0.5 8
Call For Price
HTL 5142 Optipette OP8-50 Multichannel Pipettor, Yellow, 8-Channel; 5 to 50 µL 2455172 Mfr # 5142
COMPARE
50 5 8
Call For Price
HTL 5143 Optipette OP8-200 Multichannel Pipettor, Yellow, 8-Channel; 20 to 200 µL 2455173 Mfr # 5143
COMPARE
200 20 8
Call For Price
HTL 5144 Optipette OP8-300 Multichannel Pipettor, Green, 8-Channel; 50 to 300 µL 2455174 Mfr # 5144
COMPARE
300 50 8