HTL Optipette 12-Channel Pipettors
Easily adjust ejector height
  • Ergonomic handle design
  • Easy to calibrate
  • Rotating pipetting push button prevents accidental volume change
Remarkably robust and durable in a sleek design, Optipette adjustable-volume pipettors offer optimal accuracy and precision. With a UV-resistant body and autoclavable bottom half, the soft spring system reduces pipetting forces while color-coded rings simplify volume identification. Additional spacers enable ejector height adjustment; user-calibration is easy thanks to a provided calibration key.
Call For Price
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Volume (µL) Min Volume (µL) Number Of Channels
Availability Pricing
HTL 5145 Optipette OP12-10 Multichannel Pipettor, Red, 12-Channel; 0.5 to 10 µL 2455175 Mfr # 5145
COMPARE
10 0.5 12
Call For Price
HTL 5146 Optipette OP12-50 Multichannel Pipettor, Yellow, 12-Channel; 5 to 50 µL 2455176 Mfr # 5146
COMPARE
50 5 12
Call For Price
HTL 5147 Optipette OP12-200 Multichannel Pipettor, Yellow, 12-Channel; 20 to 200 µL 2455177 Mfr # 5147
COMPARE
200 20 12
Call For Price
HTL 5148 Optipette OP12-300 Multichannel Pipettor, Green, 12-Channel; 50 to 300 µL 2455178 Mfr # 5148
COMPARE
300 50 12