Hamilton® Gastight® Cemented Needle Syringes with Conical Tip
Features a conical stainless steel (SS) needle with side port—easily penetrate septa, thin-gauged vinyls, and plastics without coring
  • Remain gas- and liquid-tight!
  • Easily handles corrosive gases and liquids, radioactive materials, and sterile solutions
  • Epoxy-cemented SS needle provides durability
  • Dead volume is limited to the internal volume of the needle
Hamilton® Gastight® syringes with cemented needle are constructed with a chemically inert glass barrel. Syringes feature a precision-machined PTFE plunger tip to create a leak-free seal, making them ideal for both liquids and gases.
Low-volume syringes include an epoxy-cemented SS needle. The "s" on the needle gauge represents a smaller inner diameter for the needle and a thicker needle wall for better durability.
£54.30 - £66.38GBP / Each
£65.16 - £79.66GBP (incl VAT)
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Needle Gauge Syringe Capacity (µL)
Availability Pricing
Hamilton Gastight® Syringe with Cemented Needle, Conical Tip, 10 µL, 26s G 0794003 Mfr # 80039
COMPARE
26s 10
£54.30GBP / Each
£65.16GBP (incl VAT)
Hamilton Gastight® Syringe with Cemented Needle, Conical Tip, 25 µL, 22s G 0794013 Mfr # 80239
COMPARE
22s 25
£60.38GBP / Each
£72.46GBP (incl VAT)
Hamilton Gastight® Syringe with Cemented Needle, Conical Tip, 50 µL, 22s G 0794021 Mfr # 80939
COMPARE
22s 50
£60.38GBP / Each
£72.46GBP (incl VAT)
Hamilton Gastight® Syringe with Cemented Needle, Conical Tip, 100 µL, 22s G 0794031 Mfr # 81039
COMPARE
22s 100
£61.19GBP / Each
£73.43GBP (incl VAT)
Hamilton Gastight® Syringe with Cemented Needle, Conical Tip, 250 µL, 22s G 0794041 Mfr # 81139
COMPARE
22s 250
£63.86GBP / Each
£76.63GBP (incl VAT)
Hamilton Gastight® Syringe with Cemented Needle, Conical Tip, 500 µL, 22 G 0794053 Mfr # 81243
COMPARE
22 500
£66.38GBP / Each
£79.66GBP (incl VAT)