GVS Neutralizing Buffer Swabs
Provides save transfer of swabbed organisms
  • Qualitative and semi-quantitative hygiene control
  • Rapid results
  • Long shelf-life
  • Sterile packed and ready-for-use
  • Individually wrapped
The neutralizing buffer swabs allow you safe transfer of swabbed organisms to the laboratory without loss in viability. The buffer inactivates the bactericidal and bacteriostatic effects of chlorine and quaternary ammonium detergents. It exhibits no toxic effects on microorganisms.
Ideal for used in the monitoring of flat or convoluted surfaces for total bacterial count.
£705.19 - £1,621.93GBP / Pkg of 125
£846.23 - £1,946.32GBP (incl VAT)
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Material Swab Total Length (InchShort) Description
Availability Pricing
GVS Neutralizing Buffer Swabs, 4mL; 125/PK 7890940 Mfr # 10498303
COMPARE
Cotton swab 3 5/64 Neutralizing Buffer Swabs, 4mL; 125/PK
£705.19GBP / Pkg of 125
£846.23GBP (incl VAT)
GVS Neutralizing Buffer Swabs, 4mL; 500/PK 7890941 Mfr # 10498304
COMPARE
Cotton swab 3 5/64 Neutralizing Buffer Swabs, 4 mL; 500/PK
£1,621.93GBP / Pkg of 500
£1,946.32GBP (incl VAT)